Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25392269-25392528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1054668 c.748C>T p.R250W Chr.4: 25392380-25392380 on Build GRCh38
1540101 c.749G>A p.R250Q Chr.4: 25392381-25392381 on Build GRCh38
4751454 c.766A>G p.T256A Chr.4: 25392398-25392398 on Build GRCh38
7161904 c.709G>C p.E237Q Chr.4: 25392341-25392341 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035052621 Chr.4: 25392494-25392494 on Build GRCh38
rs748207055 Chr.4: 25392298-25392298 on Build GRCh38
rs781603755 Chr.4: 25392461-25392461 on Build GRCh38
rs764090638 Chr.4: 25392328-25392328 on Build GRCh38
rs551684452 Chr.4: 25392426-25392426 on Build GRCh38
rs374501617 Chr.4: 25392299-25392299 on Build GRCh38
rs774477223 Chr.4: 25392332-25392332 on Build GRCh38
rs371711389 Chr.4: 25392381-25392381 on Build GRCh38
rs747256572 Chr.4: 25392306-25392306 on Build GRCh38
rs774365068 Chr.4: 25392467-25392467 on Build GRCh38
rs375380358 Chr.4: 25392469-25392469 on Build GRCh38
rs768604927 Chr.4: 25392463-25392463 on Build GRCh38
rs761368684 Chr.4: 25392339-25392339 on Build GRCh38
rs758339628 Chr.4: 25392437-25392437 on Build GRCh38
rs766016624 Chr.4: 25392380-25392380 on Build GRCh38
rs200157836 Chr.4: 25392322-25392322 on Build GRCh38
rs775341493 Chr.4: 25392310-25392310 on Build GRCh38
rs753232192 Chr.4: 25392436-25392436 on Build GRCh38
rs760362026 Chr.4: 25392358-25392358 on Build GRCh38
rs2292408 Chr.4: 25392456-25392456 on Build GRCh38
rs367748788 Chr.4: 25392355-25392355 on Build GRCh38
rs777917978 Chr.4: 25392448-25392448 on Build GRCh38
rs981757872 Chr.4: 25392314-25392314 on Build GRCh38
rs909314389 Chr.4: 25392391-25392391 on Build GRCh38
rs767060078 Chr.4: 25392342-25392342 on Build GRCh38
rs943457304 Chr.4: 25392451-25392451 on Build GRCh38
rs769413726 Chr.4: 25392309-25392309 on Build GRCh38
rs761213930 Chr.4: 25392324-25392324 on Build GRCh38
rs747116281 Chr.4: 25392452-25392452 on Build GRCh38
rs773265602 Chr.4: 25392300-25392300 on Build GRCh38
rs1049504668 Chr.4: 25392386-25392386 on Build GRCh38
rs775815190 Chr.4: 25392324-25392325 on Build GRCh38
rs375294769 Chr.4: 25392394-25392394 on Build GRCh38
rs749066990 Chr.4: 25392462-25392462 on Build GRCh38

View Full Product Details