Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18613783-18614018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM59L
Entrez Gene ID
25789
Gene Name
transmembrane protein 59 like
Gene Aliases
BSMAP, C19orf4
UniGene
Hs.329850
Gene Chromosome Location
Chr.19: 18612872-18621039 on Build GRCh38
Transcripts
NM_012109.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5133280 c.262C>T p.R88* Chr.19: 18613962-18613962 on Build GRCh38
7252468 c.311A>G p.E104G Chr.19: 18614011-18614011 on Build GRCh38
1391704 c.245G>A p.R82H Chr.19: 18613945-18613945 on Build GRCh38
6875147 c.316G>A p.A106T Chr.19: 18614016-18614016 on Build GRCh38
3403965 c.293C>T p.A98V Chr.19: 18613993-18613993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771711399 Chr.19: 18613855-18613855 on Build GRCh38
rs745961577 Chr.19: 18613846-18613846 on Build GRCh38
rs117674834 Chr.19: 18613885-18613885 on Build GRCh38
rs1016555765 Chr.19: 18613889-18613896 on Build GRCh38
rs145706240 Chr.19: 18613913-18613913 on Build GRCh38
rs1042091699 Chr.19: 18613812-18613812 on Build GRCh38
rs778716458 Chr.19: 18613851-18613851 on Build GRCh38
rs753311708 Chr.19: 18613832-18613832 on Build GRCh38
rs770778479 Chr.19: 18613942-18613942 on Build GRCh38
rs11538359 Chr.19: 18613877-18613877 on Build GRCh38
rs775097478 Chr.19: 18613950-18613950 on Build GRCh38
rs763304571 Chr.19: 18613962-18613962 on Build GRCh38
rs762266113 Chr.19: 18613971-18613971 on Build GRCh38
rs756120614 Chr.19: 18613914-18613914 on Build GRCh38
rs748161640 Chr.19: 18613854-18613854 on Build GRCh38
rs757498106 Chr.19: 18613893-18613893 on Build GRCh38
rs200515161 Chr.19: 18613901-18613901 on Build GRCh38
rs560332370 Chr.19: 18613966-18613966 on Build GRCh38
rs764513816 Chr.19: 18613889-18613889 on Build GRCh38
rs781356236 Chr.19: 18613843-18613843 on Build GRCh38
rs769135949 Chr.19: 18613946-18613946 on Build GRCh38
rs751979743 Chr.19: 18613892-18613892 on Build GRCh38
rs747991348 Chr.19: 18613925-18613925 on Build GRCh38
rs773018643 Chr.19: 18613863-18613863 on Build GRCh38
rs988473039 Chr.19: 18613974-18613974 on Build GRCh38
rs777484398 Chr.19: 18613932-18613932 on Build GRCh38
rs757586750 Chr.19: 18613839-18613839 on Build GRCh38
rs781554578 Chr.19: 18613903-18613903 on Build GRCh38
rs772046979 Chr.19: 18613931-18613931 on Build GRCh38
rs577745165 Chr.19: 18613896-18613896 on Build GRCh38
rs768039034 Chr.19: 18613990-18613990 on Build GRCh38
rs776233066 Chr.19: 18613943-18613943 on Build GRCh38
rs764712288 Chr.19: 18613963-18613963 on Build GRCh38
rs955910552 Chr.19: 18613934-18613934 on Build GRCh38
rs369319431 Chr.19: 18613867-18613867 on Build GRCh38
rs368565552 Chr.19: 18613997-18613997 on Build GRCh38
rs559064692 Chr.19: 18613945-18613945 on Build GRCh38
rs981690511 Chr.19: 18613919-18613919 on Build GRCh38
rs746790687 Chr.19: 18613935-18613935 on Build GRCh38
rs769976618 Chr.19: 18613850-18613850 on Build GRCh38
rs765646870 Chr.19: 18613827-18613827 on Build GRCh38
rs1051629150 Chr.19: 18613824-18613824 on Build GRCh38
rs760251141 Chr.19: 18613865-18613865 on Build GRCh38
rs201488046 Chr.19: 18613872-18613872 on Build GRCh38
rs372976794 Chr.19: 18613873-18613873 on Build GRCh38
rs758658360 Chr.19: 18613833-18613833 on Build GRCh38
rs745644574 Chr.19: 18613837-18613837 on Build GRCh38
rs576254642 Chr.19: 18613869-18613869 on Build GRCh38
rs749011307 Chr.19: 18613935-18613935 on Build GRCh38
rs141253088 Chr.19: 18613874-18613874 on Build GRCh38

View Full Product Details