Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 111004453-111004694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHOC2
Entrez Gene ID
8036
Gene Name
SHOC2, leucine rich repeat scaffold protein
Gene Aliases
SIAA0862, SOC2, SUR8
UniGene
Hs.104315
Gene Chromosome Location
Chr.10: 110919370-111013667 on Build GRCh38
Transcripts
NM_001269039.2 NM_001324336.1 NM_001324337.1 NM_007373.3 NR_136749.1 XM_017016702.1 XM_017016703.1 XM_017016704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1504525 c.973-1G>T p.? Chr.10: 111004605-111004605 on Build GRCh38
7276162 c.1036T>C p.Y346H Chr.10: 111004669-111004669 on Build GRCh38
465261 c.981A>G p.L327L Chr.10: 111004614-111004614 on Build GRCh38
6551322 c.1057C>T p.Q353* Chr.10: 111004690-111004690 on Build GRCh38
465262 c.996A>G p.K332K Chr.10: 111004629-111004629 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201111007 Chr.10: 111004583-111004583 on Build GRCh38
rs112013227 Chr.10: 111004515-111004515 on Build GRCh38
rs761523127 Chr.10: 111004577-111004577 on Build GRCh38
rs759167031 Chr.10: 111004593-111004594 on Build GRCh38
rs372885629 Chr.10: 111004656-111004656 on Build GRCh38
rs966407512 Chr.10: 111004662-111004662 on Build GRCh38
rs766378985 Chr.10: 111004571-111004571 on Build GRCh38
rs983654703 Chr.10: 111004549-111004549 on Build GRCh38
rs564867094 Chr.10: 111004587-111004587 on Build GRCh38
rs1033930612 Chr.10: 111004509-111004509 on Build GRCh38
rs188803618 Chr.10: 111004490-111004490 on Build GRCh38
rs1013719504 Chr.10: 111004573-111004573 on Build GRCh38
rs576005242 Chr.10: 111004579-111004579 on Build GRCh38
rs192889815 Chr.10: 111004504-111004504 on Build GRCh38
rs563914561 Chr.10: 111004576-111004576 on Build GRCh38
rs900181309 Chr.10: 111004500-111004500 on Build GRCh38
rs369588373 Chr.10: 111004508-111004508 on Build GRCh38
rs730881016 Chr.10: 111004601-111004601 on Build GRCh38
rs766924851 Chr.10: 111004611-111004611 on Build GRCh38
rs896689068 Chr.10: 111004516-111004516 on Build GRCh38
rs756466026 Chr.10: 111004599-111004599 on Build GRCh38
rs763609478 Chr.10: 111004569-111004569 on Build GRCh38

View Full Product Details