Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28111780-28112041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPO6
Entrez Gene ID
23214
Gene Name
exportin 6
Gene Aliases
EXP6, RANBP20
UniGene
Hs.460468
Gene Chromosome Location
Chr.16: 28097976-28211918 on Build GRCh38
Transcripts
NM_001270940.1 NM_015171.3 XM_005255197.3 XM_005255198.1 XM_005255199.2 XM_011545772.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
969363 c.2306C>A p.A769D Chr.16: 28111852-28111852 on Build GRCh38
6464304 c.2236G>A p.V746M Chr.16: 28111922-28111922 on Build GRCh38
6315817 c.2314C>A p.P772T Chr.16: 28111844-28111844 on Build GRCh38
6824052 c.2239C>T p.R747C Chr.16: 28111919-28111919 on Build GRCh38
4891511 c.2278C>T p.R760W Chr.16: 28111880-28111880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780913519 Chr.16: 28111914-28111914 on Build GRCh38
rs761155498 Chr.16: 28111959-28111959 on Build GRCh38
rs372203816 Chr.16: 28111871-28111871 on Build GRCh38
rs891886832 Chr.16: 28111857-28111857 on Build GRCh38
rs1054512749 Chr.16: 28111843-28111843 on Build GRCh38
rs749976444 Chr.16: 28111888-28111888 on Build GRCh38
rs759553888 Chr.16: 28111868-28111868 on Build GRCh38
rs779809302 Chr.16: 28111834-28111834 on Build GRCh38
rs11645586 Chr.16: 28111865-28111865 on Build GRCh38
rs758427183 Chr.16: 28111830-28111830 on Build GRCh38
rs765014386 Chr.16: 28112001-28112001 on Build GRCh38
rs762223020 Chr.16: 28111980-28111980 on Build GRCh38
rs367638103 Chr.16: 28111953-28111953 on Build GRCh38
rs771154737 Chr.16: 28111923-28111923 on Build GRCh38
rs545398747 Chr.16: 28111976-28111976 on Build GRCh38
rs757942295 Chr.16: 28111895-28111895 on Build GRCh38
rs201080586 Chr.16: 28111910-28111910 on Build GRCh38
rs746853825 Chr.16: 28111845-28111845 on Build GRCh38
rs761566753 Chr.16: 28111882-28111882 on Build GRCh38
rs745862947 Chr.16: 28111877-28111877 on Build GRCh38
rs145368246 Chr.16: 28111870-28111870 on Build GRCh38
rs772081390 Chr.16: 28111945-28111945 on Build GRCh38
rs751501387 Chr.16: 28111902-28111902 on Build GRCh38
rs765865185 Chr.16: 28112017-28112017 on Build GRCh38
rs768377282 Chr.16: 28111856-28111856 on Build GRCh38
rs1037316841 Chr.16: 28111972-28111972 on Build GRCh38
rs769143225 Chr.16: 28111961-28111961 on Build GRCh38
rs764928770 Chr.16: 28111887-28111887 on Build GRCh38
rs745974597 Chr.16: 28111941-28111941 on Build GRCh38
rs776466421 Chr.16: 28111866-28111866 on Build GRCh38
rs371230035 Chr.16: 28112004-28112004 on Build GRCh38
rs534087965 Chr.16: 28111904-28111904 on Build GRCh38
rs779055172 Chr.16: 28111938-28111938 on Build GRCh38
rs574144151 Chr.16: 28111867-28111867 on Build GRCh38
rs779587642 Chr.16: 28111901-28111901 on Build GRCh38
rs761210297 Chr.16: 28111878-28111878 on Build GRCh38
rs754229185 Chr.16: 28111879-28111879 on Build GRCh38
rs1003901420 Chr.16: 28111880-28111880 on Build GRCh38
rs205384 Chr.16: 28111905-28111905 on Build GRCh38
rs745315529 Chr.16: 28111806-28111806 on Build GRCh38
rs575041244 Chr.16: 28111922-28111922 on Build GRCh38

View Full Product Details