Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35003231-35003488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC4AP
Entrez Gene ID
26133
Gene Name
transient receptor potential cation channel subfamily C member 4 associated protein
Gene Aliases
C20orf188, PPP1R158, TRRP4AP, TRUSS
UniGene
Hs.168073
Gene Chromosome Location
Chr.20: 35002404-35092837 on Build GRCh38
Transcripts
NM_015638.2 NM_199368.1 XM_011528774.1 XM_017027800.1 XM_017027801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357245 c.2199G>A p.L733L Chr.20: 35003467-35003467 on Build GRCh38
723942 c.2178C>G p.L726L Chr.20: 35003488-35003488 on Build GRCh38
4535954 c.2253G>A p.E751E Chr.20: 35003413-35003413 on Build GRCh38
4981671 c.2274C>T p.F758F Chr.20: 35003266-35003266 on Build GRCh38
5955237 c.2201G>T p.R734L Chr.20: 35003465-35003465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764263935 Chr.20: 35003398-35003398 on Build GRCh38
rs1016019121 Chr.20: 35003295-35003295 on Build GRCh38
rs765387701 Chr.20: 35003311-35003311 on Build GRCh38
rs140196281 Chr.20: 35003274-35003274 on Build GRCh38
rs374852604 Chr.20: 35003386-35003386 on Build GRCh38
rs564432488 Chr.20: 35003359-35003359 on Build GRCh38
rs1002438798 Chr.20: 35003276-35003276 on Build GRCh38
rs779876351 Chr.20: 35003431-35003431 on Build GRCh38
rs748938753 Chr.20: 35003367-35003367 on Build GRCh38
rs747619622 Chr.20: 35003372-35003372 on Build GRCh38
rs973278895 Chr.20: 35003427-35003427 on Build GRCh38
rs375003906 Chr.20: 35003360-35003360 on Build GRCh38
rs755901076 Chr.20: 35003424-35003424 on Build GRCh38
rs749041992 Chr.20: 35003434-35003434 on Build GRCh38
rs763188697 Chr.20: 35003389-35003389 on Build GRCh38
rs574117291 Chr.20: 35003319-35003319 on Build GRCh38
rs894062589 Chr.20: 35003338-35003338 on Build GRCh38
rs889042420 Chr.20: 35003459-35003459 on Build GRCh38
rs768329632 Chr.20: 35003370-35003370 on Build GRCh38
rs761239995 Chr.20: 35003293-35003293 on Build GRCh38
rs777066750 Chr.20: 35003466-35003466 on Build GRCh38
rs773894861 Chr.20: 35003289-35003289 on Build GRCh38
rs759872384 Chr.20: 35003374-35003374 on Build GRCh38
rs377753023 Chr.20: 35003254-35003254 on Build GRCh38
rs777135091 Chr.20: 35003373-35003373 on Build GRCh38
rs754706800 Chr.20: 35003440-35003440 on Build GRCh38
rs11481073 Chr.20: 35003394-35003395 on Build GRCh38
rs769338101 Chr.20: 35003256-35003256 on Build GRCh38
rs781120142 Chr.20: 35003333-35003333 on Build GRCh38
rs771594974 Chr.20: 35003465-35003465 on Build GRCh38
rs981797154 Chr.20: 35003353-35003353 on Build GRCh38
rs368271092 Chr.20: 35003410-35003410 on Build GRCh38
rs970822375 Chr.20: 35003342-35003342 on Build GRCh38
rs751592821 Chr.20: 35003322-35003322 on Build GRCh38
rs552619241 Chr.20: 35003358-35003358 on Build GRCh38
rs59909520 Chr.20: 35003304-35003305 on Build GRCh38
rs769619617 Chr.20: 35003365-35003365 on Build GRCh38
rs777811744 Chr.20: 35003318-35003318 on Build GRCh38
rs775150907 Chr.20: 35003270-35003270 on Build GRCh38
rs758543253 Chr.20: 35003305-35003305 on Build GRCh38
rs878971863 Chr.20: 35003341-35003341 on Build GRCh38
rs977939719 Chr.20: 35003394-35003394 on Build GRCh38
rs766941277 Chr.20: 35003307-35003307 on Build GRCh38
rs924703070 Chr.20: 35003409-35003409 on Build GRCh38
rs397963996 Chr.20: 35003310-35003311 on Build GRCh38
rs370371907 Chr.20: 35003371-35003371 on Build GRCh38
rs759875405 Chr.20: 35003310-35003310 on Build GRCh38
rs397757541 Chr.20: 35003397-35003398 on Build GRCh38
rs370295838 Chr.20: 35003330-35003330 on Build GRCh38
rs752947568 Chr.20: 35003312-35003312 on Build GRCh38
rs376209935 Chr.20: 35003313-35003313 on Build GRCh38
rs532450467 Chr.20: 35003366-35003366 on Build GRCh38
rs370841577 Chr.20: 35003446-35003446 on Build GRCh38
rs754182997 Chr.20: 35003309-35003309 on Build GRCh38
rs778639752 Chr.20: 35003443-35003443 on Build GRCh38
rs750371131 Chr.20: 35003416-35003416 on Build GRCh38
rs757271512 Chr.20: 35003400-35003400 on Build GRCh38

View Full Product Details