Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70293499-70293772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPG
Entrez Gene ID
6637
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
SMG, Sm-G
UniGene
Hs.465167
Gene Chromosome Location
Chr.2: 70281362-70293771 on Build GRCh38
Transcripts
NM_001317165.1 NM_001317166.1 NM_001317171.1 NM_003096.3 XM_017004774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839903 c.3G>A p.M1I Chr.2: 70293647-70293647 on Build GRCh38
4095342 c.10G>T p.A4S Chr.2: 70293640-70293640 on Build GRCh38
1022516 c.14A>G p.H5R Chr.2: 70293636-70293636 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780871842 Chr.2: 70293683-70293683 on Build GRCh38
rs775954821 Chr.2: 70293651-70293651 on Build GRCh38
rs15656 Chr.2: 70293714-70293714 on Build GRCh38
rs370473703 Chr.2: 70293691-70293691 on Build GRCh38
rs536568847 Chr.2: 70293750-70293750 on Build GRCh38
rs776461879 Chr.2: 70293626-70293626 on Build GRCh38
rs767879369 Chr.2: 70293611-70293612 on Build GRCh38
rs769221300 Chr.2: 70293653-70293653 on Build GRCh38
rs1044916424 Chr.2: 70293747-70293747 on Build GRCh38
rs561201852 Chr.2: 70293580-70293580 on Build GRCh38
rs965064512 Chr.2: 70293708-70293708 on Build GRCh38
rs906515942 Chr.2: 70293539-70293539 on Build GRCh38
rs769520794 Chr.2: 70293650-70293650 on Build GRCh38
rs1045345339 Chr.2: 70293550-70293550 on Build GRCh38
rs376553694 Chr.2: 70293715-70293715 on Build GRCh38
rs540402095 Chr.2: 70293686-70293686 on Build GRCh38
rs373447773 Chr.2: 70293696-70293696 on Build GRCh38
rs756875894 Chr.2: 70293679-70293679 on Build GRCh38
rs910028055 Chr.2: 70293732-70293732 on Build GRCh38
rs760913250 Chr.2: 70293666-70293666 on Build GRCh38
rs34968038 Chr.2: 70293661-70293662 on Build GRCh38
rs766441729 Chr.2: 70293670-70293670 on Build GRCh38
rs752545398 Chr.2: 70293674-70293674 on Build GRCh38
rs763792385 Chr.2: 70293697-70293697 on Build GRCh38
rs745492903 Chr.2: 70293632-70293632 on Build GRCh38
rs375246588 Chr.2: 70293660-70293660 on Build GRCh38
rs754811913 Chr.2: 70293575-70293575 on Build GRCh38
rs918260985 Chr.2: 70293712-70293712 on Build GRCh38
rs758356324 Chr.2: 70293602-70293602 on Build GRCh38
rs373366542 Chr.2: 70293571-70293571 on Build GRCh38
rs377108614 Chr.2: 70293676-70293676 on Build GRCh38
rs372874888 Chr.2: 70293662-70293662 on Build GRCh38
rs891674502 Chr.2: 70293664-70293664 on Build GRCh38
rs763161158 Chr.2: 70293652-70293652 on Build GRCh38
rs905125530 Chr.2: 70293573-70293573 on Build GRCh38
rs948243782 Chr.2: 70293734-70293734 on Build GRCh38
rs188770074 Chr.2: 70293547-70293547 on Build GRCh38
rs376253508 Chr.2: 70293615-70293615 on Build GRCh38
rs1034712471 Chr.2: 70293629-70293629 on Build GRCh38
rs745331803 Chr.2: 70293685-70293685 on Build GRCh38
rs758112405 Chr.2: 70293675-70293675 on Build GRCh38
rs201792473 Chr.2: 70293579-70293579 on Build GRCh38
rs185436321 Chr.2: 70293716-70293716 on Build GRCh38
rs1003589227 Chr.2: 70293574-70293574 on Build GRCh38
rs866948523 Chr.2: 70293553-70293553 on Build GRCh38
rs1045576335 Chr.2: 70293525-70293525 on Build GRCh38
rs973600434 Chr.2: 70293682-70293682 on Build GRCh38
rs976455711 Chr.2: 70293710-70293710 on Build GRCh38
rs777511494 Chr.2: 70293605-70293605 on Build GRCh38
rs75215614 Chr.2: 70293719-70293719 on Build GRCh38
rs770559448 Chr.2: 70293625-70293625 on Build GRCh38
rs747853720 Chr.2: 70293593-70293593 on Build GRCh38
rs374092476 Chr.2: 70293705-70293705 on Build GRCh38
rs1052203098 Chr.2: 70293523-70293523 on Build GRCh38
rs750698198 Chr.2: 70293665-70293665 on Build GRCh38
rs370306808 Chr.2: 70293542-70293542 on Build GRCh38
rs191710102 Chr.2: 70293695-70293695 on Build GRCh38
rs1021384283 Chr.2: 70293689-70293689 on Build GRCh38
rs4567961 Chr.2: 70293743-70293743 on Build GRCh38
rs560407173 Chr.2: 70293692-70293692 on Build GRCh38

View Full Product Details