Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50481708-50481955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC10
Entrez Gene ID
284361
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19orf63, HSM1, HSS1
UniGene
Hs.448941
Gene Chromosome Location
Chr.19: 50476477-50505905 on Build GRCh38
Transcripts
NM_175063.5 NM_206538.3 XM_006723163.3 XM_011526818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390418 c.691C>T p.L231L Chr.19: 50481873-50481873 on Build GRCh38
292630 c.692_693insG p.A234fs*56 Chr.19: 50481874-50481875 on Build GRCh38
6230140 c.749C>T p.P250L Chr.19: 50481931-50481931 on Build GRCh38
1579655 c.693delG p.A234fs*>21 Chr.19: 50481875-50481875 on Build GRCh38
6653611 c.701C>G p.A234G Chr.19: 50481883-50481883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754075492 Chr.19: 50481875-50481875 on Build GRCh38
rs530683575 Chr.19: 50481785-50481785 on Build GRCh38
rs754547061 Chr.19: 50481895-50481895 on Build GRCh38
rs1022707601 Chr.19: 50481806-50481806 on Build GRCh38
rs1055887719 Chr.19: 50481782-50481782 on Build GRCh38
rs1726783 Chr.19: 50481857-50481857 on Build GRCh38
rs1016868432 Chr.19: 50481877-50481877 on Build GRCh38
rs549810670 Chr.19: 50481819-50481819 on Build GRCh38
rs771424054 Chr.19: 50481828-50481828 on Build GRCh38
rs901735433 Chr.19: 50481767-50481767 on Build GRCh38
rs779918120 Chr.19: 50481883-50481883 on Build GRCh38
rs993308436 Chr.19: 50481768-50481768 on Build GRCh38
rs761982110 Chr.19: 50481876-50481876 on Build GRCh38
rs12609351 Chr.19: 50481781-50481781 on Build GRCh38
rs549338223 Chr.19: 50481803-50481803 on Build GRCh38
rs764147364 Chr.19: 50481860-50481860 on Build GRCh38
rs970160616 Chr.19: 50481817-50481817 on Build GRCh38
rs200206091 Chr.19: 50481874-50481875 on Build GRCh38
rs747835919 Chr.19: 50481904-50481904 on Build GRCh38
rs775920286 Chr.19: 50481924-50481924 on Build GRCh38
rs571186845 Chr.19: 50481879-50481879 on Build GRCh38
rs575791499 Chr.19: 50481914-50481914 on Build GRCh38
rs762710427 Chr.19: 50481936-50481936 on Build GRCh38
rs887604810 Chr.19: 50481786-50481786 on Build GRCh38
rs144996610 Chr.19: 50481913-50481913 on Build GRCh38
rs750653782 Chr.19: 50481933-50481933 on Build GRCh38
rs765037624 Chr.19: 50481878-50481878 on Build GRCh38
rs946380476 Chr.19: 50481747-50481747 on Build GRCh38
rs375521469 Chr.19: 50481926-50481926 on Build GRCh38
rs751498488 Chr.19: 50481885-50481885 on Build GRCh38
rs955174997 Chr.19: 50481891-50481891 on Build GRCh38
rs199946407 Chr.19: 50481925-50481925 on Build GRCh38
rs1006406692 Chr.19: 50481793-50481793 on Build GRCh38
rs764239506 Chr.19: 50481875-50481875 on Build GRCh38
rs926174844 Chr.19: 50481780-50481780 on Build GRCh38
rs760114642 Chr.19: 50481842-50481842 on Build GRCh38
rs370195879 Chr.19: 50481931-50481931 on Build GRCh38
rs746263716 Chr.19: 50481917-50481917 on Build GRCh38
rs371502898 Chr.19: 50481886-50481886 on Build GRCh38
rs868186123 Chr.19: 50481844-50481844 on Build GRCh38
rs200797859 Chr.19: 50481882-50481882 on Build GRCh38
rs373371781 Chr.19: 50481932-50481932 on Build GRCh38
rs534628081 Chr.19: 50481835-50481835 on Build GRCh38
rs772509211 Chr.19: 50481843-50481843 on Build GRCh38
rs866028566 Chr.19: 50481758-50481758 on Build GRCh38
rs776034405 Chr.19: 50481853-50481853 on Build GRCh38
rs767952336 Chr.19: 50481877-50481878 on Build GRCh38
rs567561385 Chr.19: 50481820-50481820 on Build GRCh38
rs943382273 Chr.19: 50481905-50481905 on Build GRCh38
rs200196256 Chr.19: 50481769-50481769 on Build GRCh38
rs910630135 Chr.19: 50481902-50481902 on Build GRCh38
rs776642182 Chr.19: 50481927-50481927 on Build GRCh38
rs780779977 Chr.19: 50481903-50481903 on Build GRCh38
rs57237609 Chr.19: 50481923-50481923 on Build GRCh38

View Full Product Details