Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121923777-121924048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR66
Entrez Gene ID
144406
Gene Name
WD repeat domain 66
Gene Aliases
CaM-IP4
UniGene
Hs.709837
Gene Chromosome Location
Chr.12: 121918557-122003927 on Build GRCh38
Transcripts
NM_001178003.1 NM_144668.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5803628 c.551A>T p.E184V Chr.12: 121923794-121923794 on Build GRCh38
1359531 c.616_617delAG p.E207fs*35 Chr.12: 121923859-121923860 on Build GRCh38
1359530 c.579A>T p.E193D Chr.12: 121923822-121923822 on Build GRCh38
3457232 c.720G>A p.K240K Chr.12: 121923963-121923963 on Build GRCh38
936126 c.726A>C p.K242N Chr.12: 121923969-121923969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375756651 Chr.12: 121923801-121923801 on Build GRCh38
rs754523723 Chr.12: 121923974-121923974 on Build GRCh38
rs894654214 Chr.12: 121923818-121923818 on Build GRCh38
rs371486114 Chr.12: 121923977-121923977 on Build GRCh38
rs771922304 Chr.12: 121923881-121923881 on Build GRCh38
rs921145061 Chr.12: 121923987-121923987 on Build GRCh38
rs753085705 Chr.12: 121923830-121923830 on Build GRCh38
rs759710570 Chr.12: 121923999-121923999 on Build GRCh38
rs754046262 Chr.12: 121923806-121923806 on Build GRCh38
rs377324406 Chr.12: 121923871-121923871 on Build GRCh38
rs759735665 Chr.12: 121923892-121923892 on Build GRCh38
rs375556697 Chr.12: 121923934-121923934 on Build GRCh38
rs761482571 Chr.12: 121923878-121923878 on Build GRCh38
rs770968035 Chr.12: 121924003-121924003 on Build GRCh38
rs34885077 Chr.12: 121924024-121924024 on Build GRCh38
rs758833653 Chr.12: 121923833-121923833 on Build GRCh38
rs765520190 Chr.12: 121923899-121923899 on Build GRCh38
rs1720071 Chr.12: 121923898-121923898 on Build GRCh38
rs760622578 Chr.12: 121923802-121923802 on Build GRCh38
rs746834273 Chr.12: 121924002-121924002 on Build GRCh38
rs199861707 Chr.12: 121923933-121923933 on Build GRCh38
rs755243380 Chr.12: 121923814-121923814 on Build GRCh38
rs747577416 Chr.12: 121923843-121923843 on Build GRCh38
rs771695038 Chr.12: 121923978-121923978 on Build GRCh38
rs776745573 Chr.12: 121924012-121924012 on Build GRCh38
rs764536808 Chr.12: 121923910-121923910 on Build GRCh38
rs748865361 Chr.12: 121923868-121923868 on Build GRCh38
rs772964725 Chr.12: 121923882-121923882 on Build GRCh38
rs925665642 Chr.12: 121924022-121924022 on Build GRCh38
rs375750046 Chr.12: 121923916-121923916 on Build GRCh38
rs368528812 Chr.12: 121923965-121923965 on Build GRCh38
rs987635127 Chr.12: 121923907-121923907 on Build GRCh38
rs758644182 Chr.12: 121923903-121923903 on Build GRCh38
rs868055135 Chr.12: 121923973-121923973 on Build GRCh38
rs778105551 Chr.12: 121923837-121923837 on Build GRCh38
rs769129905 Chr.12: 121923849-121923849 on Build GRCh38
rs978054287 Chr.12: 121923989-121923989 on Build GRCh38
rs180719819 Chr.12: 121923815-121923815 on Build GRCh38
rs150940900 Chr.12: 121923917-121923917 on Build GRCh38
rs34703321 Chr.12: 121923805-121923805 on Build GRCh38
rs952577194 Chr.12: 121924013-121924014 on Build GRCh38
rs774265217 Chr.12: 121923948-121923949 on Build GRCh38
rs201116442 Chr.12: 121923888-121923888 on Build GRCh38
rs774039492 Chr.12: 121923874-121923874 on Build GRCh38
rs779332951 Chr.12: 121923816-121923816 on Build GRCh38
rs200006803 Chr.12: 121923889-121923889 on Build GRCh38
rs773053323 Chr.12: 121923986-121923986 on Build GRCh38
rs1019090843 Chr.12: 121923932-121923932 on Build GRCh38
rs397823191 Chr.12: 121924027-121924027 on Build GRCh38
rs373280656 Chr.12: 121923864-121923864 on Build GRCh38
rs370548434 Chr.12: 121923879-121923879 on Build GRCh38

View Full Product Details