Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48980932-48981199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT1
Entrez Gene ID
7471
Gene Name
Wnt family member 1
Gene Aliases
BMND16, INT1, OI15
UniGene
Hs.248164
Gene Chromosome Location
Chr.12: 48978453-48982613 on Build GRCh38
Transcripts
NM_005430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439260 c.633C>A p.F211L Chr.12: 48981160-48981160 on Build GRCh38
4930096 c.670T>C p.S224P Chr.12: 48981197-48981197 on Build GRCh38
6823157 c.637G>A p.E213K Chr.12: 48981164-48981164 on Build GRCh38
5519141 c.625-7C>A p.? Chr.12: 48981145-48981145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928181226 Chr.12: 48981006-48981006 on Build GRCh38
rs778947109 Chr.12: 48981113-48981113 on Build GRCh38
rs372456602 Chr.12: 48981125-48981125 on Build GRCh38
rs959396832 Chr.12: 48980959-48980959 on Build GRCh38
rs756140371 Chr.12: 48981110-48981110 on Build GRCh38
rs535491916 Chr.12: 48981058-48981058 on Build GRCh38
rs777059195 Chr.12: 48981145-48981145 on Build GRCh38
rs990969395 Chr.12: 48980986-48980986 on Build GRCh38
rs932063569 Chr.12: 48981118-48981118 on Build GRCh38
rs397514702 Chr.12: 48981179-48981179 on Build GRCh38
rs771142889 Chr.12: 48981137-48981137 on Build GRCh38
rs746357834 Chr.12: 48981154-48981154 on Build GRCh38
rs968283620 Chr.12: 48981023-48981023 on Build GRCh38
rs770483597 Chr.12: 48981170-48981170 on Build GRCh38
rs777930556 Chr.12: 48981123-48981123 on Build GRCh38
rs970735199 Chr.12: 48981085-48981085 on Build GRCh38
rs771501280 Chr.12: 48981074-48981074 on Build GRCh38
rs201651278 Chr.12: 48981144-48981144 on Build GRCh38
rs199735570 Chr.12: 48981171-48981171 on Build GRCh38
rs959282870 Chr.12: 48981041-48981041 on Build GRCh38
rs373208530 Chr.12: 48981052-48981052 on Build GRCh38
rs576120618 Chr.12: 48981014-48981014 on Build GRCh38
rs747423134 Chr.12: 48981008-48981008 on Build GRCh38
rs1040369122 Chr.12: 48981093-48981093 on Build GRCh38
rs944690130 Chr.12: 48981078-48981078 on Build GRCh38
rs748272929 Chr.12: 48981122-48981122 on Build GRCh38
rs746764049 Chr.12: 48981105-48981106 on Build GRCh38
rs922008435 Chr.12: 48981114-48981114 on Build GRCh38
rs997699695 Chr.12: 48980952-48980952 on Build GRCh38
rs982677886 Chr.12: 48980987-48980987 on Build GRCh38
rs556287136 Chr.12: 48980980-48980980 on Build GRCh38
rs950562275 Chr.12: 48980970-48980970 on Build GRCh38
rs192013975 Chr.12: 48981071-48981071 on Build GRCh38
rs540489736 Chr.12: 48981182-48981182 on Build GRCh38
rs763732264 Chr.12: 48981181-48981181 on Build GRCh38

View Full Product Details