Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15074098-15074357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBSN
Entrez Gene ID
64145
Gene Name
rabenosyn, RAB effector
Gene Aliases
Rabenosyn-5, ZFYVE20
UniGene
Hs.475565
Gene Chromosome Location
Chr.3: 15070069-15099163 on Build GRCh38
Transcripts
NM_001302378.1 NM_022340.3 XM_005265384.4 XM_005265385.4 XM_011534001.2 XM_017007023.1 XM_017007024.1 XM_017007025.1 XM_017007026.1 XM_017007027.1 XM_017007028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6826980 c.2005G>A p.E669K Chr.3: 15074132-15074132 on Build GRCh38
3846519 c.2021T>G p.V674G Chr.3: 15074116-15074116 on Build GRCh38
1731072 c.1816G>T p.V606F Chr.3: 15074321-15074321 on Build GRCh38
5863765 c.1966C>T p.R656C Chr.3: 15074171-15074171 on Build GRCh38
3729230 c.1942G>T p.G648W Chr.3: 15074195-15074195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569450225 Chr.3: 15074152-15074152 on Build GRCh38
rs768820778 Chr.3: 15074236-15074236 on Build GRCh38
rs766897022 Chr.3: 15074322-15074322 on Build GRCh38
rs111361361 Chr.3: 15074156-15074156 on Build GRCh38
rs754469602 Chr.3: 15074190-15074190 on Build GRCh38
rs375709820 Chr.3: 15074126-15074126 on Build GRCh38
rs977022283 Chr.3: 15074136-15074136 on Build GRCh38
rs908728910 Chr.3: 15074321-15074321 on Build GRCh38
rs1045610590 Chr.3: 15074275-15074275 on Build GRCh38
rs751197202 Chr.3: 15074309-15074309 on Build GRCh38
rs745964237 Chr.3: 15074260-15074260 on Build GRCh38
rs776843735 Chr.3: 15074291-15074291 on Build GRCh38
rs949908294 Chr.3: 15074232-15074232 on Build GRCh38
rs148558728 Chr.3: 15074162-15074162 on Build GRCh38
rs747986773 Chr.3: 15074204-15074204 on Build GRCh38
rs755978451 Chr.3: 15074207-15074207 on Build GRCh38
rs566679794 Chr.3: 15074308-15074308 on Build GRCh38
rs750126292 Chr.3: 15074160-15074160 on Build GRCh38
rs991849833 Chr.3: 15074281-15074281 on Build GRCh38
rs764563601 Chr.3: 15074273-15074273 on Build GRCh38
rs754557505 Chr.3: 15074311-15074311 on Build GRCh38
rs552182343 Chr.3: 15074151-15074151 on Build GRCh38
rs147577833 Chr.3: 15074170-15074170 on Build GRCh38
rs373403772 Chr.3: 15074145-15074145 on Build GRCh38
rs113330704 Chr.3: 15074155-15074155 on Build GRCh38
rs760103783 Chr.3: 15074144-15074144 on Build GRCh38
rs777408454 Chr.3: 15074209-15074209 on Build GRCh38
rs918433064 Chr.3: 15074230-15074230 on Build GRCh38
rs905821751 Chr.3: 15074304-15074304 on Build GRCh38
rs368892679 Chr.3: 15074132-15074132 on Build GRCh38
rs772001321 Chr.3: 15074268-15074268 on Build GRCh38
rs751107131 Chr.3: 15074171-15074171 on Build GRCh38
rs1018813001 Chr.3: 15074146-15074146 on Build GRCh38
rs9851219 Chr.3: 15074216-15074216 on Build GRCh38
rs139954695 Chr.3: 15074130-15074130 on Build GRCh38
rs966960752 Chr.3: 15074133-15074133 on Build GRCh38
rs776099752 Chr.3: 15074270-15074270 on Build GRCh38
rs941449139 Chr.3: 15074208-15074208 on Build GRCh38
rs771138617 Chr.3: 15074218-15074218 on Build GRCh38
rs774492339 Chr.3: 15074229-15074229 on Build GRCh38
rs144799723 Chr.3: 15074153-15074153 on Build GRCh38
rs202109121 Chr.3: 15074192-15074192 on Build GRCh38
rs550571960 Chr.3: 15074122-15074124 on Build GRCh38
rs762757915 Chr.3: 15074298-15074298 on Build GRCh38
rs761051401 Chr.3: 15074271-15074271 on Build GRCh38

View Full Product Details