Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26041619-26041866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A2
Entrez Gene ID
7780
Gene Name
solute carrier family 30 member 2
Gene Aliases
PP12488, TNZD, ZNT2, ZnT-2
UniGene
Hs.143545
Gene Chromosome Location
Chr.1: 26038022-26046138 on Build GRCh38
Transcripts
NM_001004434.2 NM_032513.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5992835 c.757A>C p.I253L Chr.1: 26041781-26041781 on Build GRCh38
4856611 c.756C>T p.P252P Chr.1: 26041782-26041782 on Build GRCh38
5502184 c.791G>T p.G264V Chr.1: 26041747-26041747 on Build GRCh38
1320240 c.770T>G p.V257G Chr.1: 26041768-26041768 on Build GRCh38
907571 c.790G>T p.G264W Chr.1: 26041748-26041748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746722376 Chr.1: 26041703-26041703 on Build GRCh38
rs770233361 Chr.1: 26041731-26041731 on Build GRCh38
rs764921143 Chr.1: 26041790-26041790 on Build GRCh38
rs537744338 Chr.1: 26041651-26041651 on Build GRCh38
rs372502581 Chr.1: 26041657-26041657 on Build GRCh38
rs777996663 Chr.1: 26041828-26041828 on Build GRCh38
rs778874275 Chr.1: 26041659-26041659 on Build GRCh38
rs750412254 Chr.1: 26041794-26041794 on Build GRCh38
rs773717920 Chr.1: 26041732-26041732 on Build GRCh38
rs761623094 Chr.1: 26041776-26041776 on Build GRCh38
rs748457509 Chr.1: 26041722-26041722 on Build GRCh38
rs767921934 Chr.1: 26041812-26041812 on Build GRCh38
rs1039156451 Chr.1: 26041840-26041840 on Build GRCh38
rs757978172 Chr.1: 26041678-26041678 on Build GRCh38
rs368895984 Chr.1: 26041654-26041654 on Build GRCh38
rs749926872 Chr.1: 26041663-26041663 on Build GRCh38
rs1316852 Chr.1: 26041770-26041770 on Build GRCh38
rs368388103 Chr.1: 26041813-26041813 on Build GRCh38
rs759768447 Chr.1: 26041763-26041763 on Build GRCh38
rs749054301 Chr.1: 26041650-26041650 on Build GRCh38
rs372442248 Chr.1: 26041769-26041769 on Build GRCh38
rs780951720 Chr.1: 26041815-26041815 on Build GRCh38
rs779397788 Chr.1: 26041684-26041684 on Build GRCh38
rs972394706 Chr.1: 26041658-26041658 on Build GRCh38
rs138246170 Chr.1: 26041739-26041739 on Build GRCh38
rs765967769 Chr.1: 26041804-26041804 on Build GRCh38
rs756549333 Chr.1: 26041825-26041825 on Build GRCh38
rs370189147 Chr.1: 26041656-26041656 on Build GRCh38
rs570344520 Chr.1: 26041720-26041720 on Build GRCh38
rs748543540 Chr.1: 26041819-26041819 on Build GRCh38
rs758261486 Chr.1: 26041802-26041802 on Build GRCh38
rs901953386 Chr.1: 26041842-26041842 on Build GRCh38
rs202003978 Chr.1: 26041758-26041758 on Build GRCh38
rs763339514 Chr.1: 26041740-26041740 on Build GRCh38
rs768406992 Chr.1: 26041709-26041709 on Build GRCh38
rs1049692403 Chr.1: 26041764-26041764 on Build GRCh38
rs770326347 Chr.1: 26041655-26041655 on Build GRCh38
rs781697562 Chr.1: 26041716-26041716 on Build GRCh38

View Full Product Details