Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64554823-64555048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMURF2
Entrez Gene ID
64750
Gene Name
SMAD specific E3 ubiquitin protein ligase 2
Gene Aliases
-
UniGene
Hs.515011
Gene Chromosome Location
Chr.17: 64542282-64662268 on Build GRCh38
Transcripts
NM_022739.3 XM_005257585.3 XM_011525128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820265 c.1678A>G p.I560V Chr.17: 64554926-64554926 on Build GRCh38
5001563 c.1614G>T p.E538D Chr.17: 64554990-64554990 on Build GRCh38
4068742 c.1716T>C p.P572P Chr.17: 64554888-64554888 on Build GRCh38
6349392 c.1723G>T p.E575* Chr.17: 64554881-64554881 on Build GRCh38
7391157 c.1643C>T p.T548I Chr.17: 64554961-64554961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973078000 Chr.17: 64554848-64554848 on Build GRCh38
rs781942844 Chr.17: 64554943-64554944 on Build GRCh38
rs533982244 Chr.17: 64554864-64554864 on Build GRCh38
rs782154487 Chr.17: 64554982-64554982 on Build GRCh38
rs376057787 Chr.17: 64554890-64554890 on Build GRCh38
rs370520140 Chr.17: 64554961-64554961 on Build GRCh38
rs782439202 Chr.17: 64555018-64555018 on Build GRCh38
rs920881297 Chr.17: 64554851-64554851 on Build GRCh38
rs781801641 Chr.17: 64555006-64555006 on Build GRCh38
rs373923799 Chr.17: 64554965-64554965 on Build GRCh38
rs782802096 Chr.17: 64554996-64554996 on Build GRCh38
rs553108922 Chr.17: 64555010-64555010 on Build GRCh38
rs782254986 Chr.17: 64554862-64554862 on Build GRCh38
rs782607325 Chr.17: 64554858-64554858 on Build GRCh38
rs912073687 Chr.17: 64554991-64554991 on Build GRCh38
rs782068855 Chr.17: 64554898-64554898 on Build GRCh38
rs781850491 Chr.17: 64554977-64554977 on Build GRCh38
rs782311363 Chr.17: 64554904-64554904 on Build GRCh38
rs782776343 Chr.17: 64554971-64554971 on Build GRCh38

View Full Product Details