Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9873599-9873860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHROOM2
Entrez Gene ID
357
Gene Name
shroom family member 2
Gene Aliases
APXL, HSAPXL
UniGene
Hs.567236
Gene Chromosome Location
Chr.X: 9786406-9949443 on Build GRCh38
Transcripts
NM_001649.3 XM_005274500.4

SNP Detail

SNP ID Chromosome Location
rs909427616 Chr.X: 9873723-9873723 on Build GRCh38
rs201837863 Chr.X: 9873708-9873708 on Build GRCh38
rs781044272 Chr.X: 9873779-9873779 on Build GRCh38
rs758297863 Chr.X: 9873637-9873637 on Build GRCh38
rs372682515 Chr.X: 9873829-9873829 on Build GRCh38
rs149912209 Chr.X: 9873681-9873681 on Build GRCh38
rs145000598 Chr.X: 9873683-9873683 on Build GRCh38
rs761019672 Chr.X: 9873686-9873686 on Build GRCh38
rs201114383 Chr.X: 9873674-9873674 on Build GRCh38
rs779929341 Chr.X: 9873638-9873638 on Build GRCh38
rs146757558 Chr.X: 9873753-9873753 on Build GRCh38
rs139542964 Chr.X: 9873678-9873678 on Build GRCh38
rs202237359 Chr.X: 9873835-9873835 on Build GRCh38
rs138116190 Chr.X: 9873834-9873834 on Build GRCh38
rs778904040 Chr.X: 9873808-9873808 on Build GRCh38
rs758068903 Chr.X: 9873738-9873738 on Build GRCh38
rs868696257 Chr.X: 9873757-9873757 on Build GRCh38
rs750103047 Chr.X: 9873711-9873711 on Build GRCh38
rs755574980 Chr.X: 9873789-9873789 on Build GRCh38
rs368631870 Chr.X: 9873813-9873813 on Build GRCh38
rs1047996391 Chr.X: 9873712-9873712 on Build GRCh38
rs751468736 Chr.X: 9873759-9873759 on Build GRCh38
rs374833426 Chr.X: 9873672-9873672 on Build GRCh38
rs750045862 Chr.X: 9873623-9873623 on Build GRCh38
rs771024837 Chr.X: 9873677-9873677 on Build GRCh38
rs201633094 Chr.X: 9873675-9873675 on Build GRCh38
rs142199040 Chr.X: 9873709-9873709 on Build GRCh38
rs200846360 Chr.X: 9873724-9873724 on Build GRCh38
rs777173762 Chr.X: 9873699-9873699 on Build GRCh38
rs199860180 Chr.X: 9873652-9873652 on Build GRCh38
rs772625017 Chr.X: 9873682-9873682 on Build GRCh38
rs764891545 Chr.X: 9873783-9873783 on Build GRCh38
rs908800096 Chr.X: 9873825-9873825 on Build GRCh38
rs140479341 Chr.X: 9873795-9873795 on Build GRCh38
rs371735679 Chr.X: 9873836-9873836 on Build GRCh38
rs879221119 Chr.X: 9873687-9873687 on Build GRCh38
rs149096987 Chr.X: 9873694-9873694 on Build GRCh38
rs754772424 Chr.X: 9873771-9873771 on Build GRCh38

View Full Product Details