Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53082543-53082772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPYL2
Entrez Gene ID
64061
Gene Name
TSPY like 2
Gene Aliases
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
UniGene
Hs.136164
Gene Chromosome Location
Chr.X: 53082327-53088546 on Build GRCh38
Transcripts
NM_022117.3 XM_006724592.3 XM_017029726.1 XM_017029727.1

SNP Detail

SNP ID Chromosome Location
rs781973800 Chr.X: 53082721-53082721 on Build GRCh38
rs782335358 Chr.X: 53082684-53082684 on Build GRCh38
rs781871680 Chr.X: 53082566-53082566 on Build GRCh38
rs782084626 Chr.X: 53082735-53082735 on Build GRCh38
rs782451065 Chr.X: 53082628-53082628 on Build GRCh38
rs1002926853 Chr.X: 53082587-53082587 on Build GRCh38
rs782544701 Chr.X: 53082676-53082676 on Build GRCh38
rs887205918 Chr.X: 53082582-53082582 on Build GRCh38
rs782760564 Chr.X: 53082712-53082712 on Build GRCh38
rs782748092 Chr.X: 53082588-53082588 on Build GRCh38
rs781793525 Chr.X: 53082748-53082748 on Build GRCh38
rs1025619221 Chr.X: 53082576-53082576 on Build GRCh38
rs782223791 Chr.X: 53082683-53082683 on Build GRCh38
rs781884842 Chr.X: 53082576-53082577 on Build GRCh38
rs782126143 Chr.X: 53082711-53082711 on Build GRCh38
rs782658165 Chr.X: 53082682-53082682 on Build GRCh38
rs782171306 Chr.X: 53082694-53082694 on Build GRCh38
rs782014870 Chr.X: 53082708-53082708 on Build GRCh38
rs782294711 Chr.X: 53082660-53082660 on Build GRCh38
rs782504396 Chr.X: 53082572-53082572 on Build GRCh38
rs781932166 Chr.X: 53082687-53082687 on Build GRCh38
rs782582177 Chr.X: 53082636-53082636 on Build GRCh38
rs781823840 Chr.X: 53082606-53082606 on Build GRCh38
rs782412723 Chr.X: 53082706-53082706 on Build GRCh38

View Full Product Details