Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44255248-44255499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35B2
Entrez Gene ID
347734
Gene Name
solute carrier family 35 member B2
Gene Aliases
PAPST1, SLL, UGTrel4
UniGene
Hs.182885
Gene Chromosome Location
Chr.6: 44254101-44257890 on Build GRCh38
Transcripts
NM_001286509.1 NM_001286510.1 NM_001286511.1 NM_001286512.1 NM_001286513.1 NM_001286517.1 NM_001286519.1 NM_001286520.1 NM_178148.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116159 c.692C>G p.S231C Chr.6: 44255313-44255313 on Build GRCh38
6781635 c.604G>A p.E202K Chr.6: 44255401-44255401 on Build GRCh38
5964445 c.533G>A p.R178Q Chr.6: 44255472-44255472 on Build GRCh38
4913493 c.705C>T p.Y235Y Chr.6: 44255300-44255300 on Build GRCh38
1079744 c.686T>G p.L229R Chr.6: 44255319-44255319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747186898 Chr.6: 44255325-44255325 on Build GRCh38
rs765716878 Chr.6: 44255375-44255375 on Build GRCh38
rs182428561 Chr.6: 44255360-44255360 on Build GRCh38
rs763894101 Chr.6: 44255354-44255354 on Build GRCh38
rs762828763 Chr.6: 44255339-44255339 on Build GRCh38
rs201227721 Chr.6: 44255279-44255279 on Build GRCh38
rs745850555 Chr.6: 44255335-44255335 on Build GRCh38
rs777869514 Chr.6: 44255317-44255317 on Build GRCh38
rs966448911 Chr.6: 44255342-44255342 on Build GRCh38
rs758675008 Chr.6: 44255311-44255311 on Build GRCh38
rs538318523 Chr.6: 44255300-44255300 on Build GRCh38
rs769912444 Chr.6: 44255457-44255457 on Build GRCh38
rs779286571 Chr.6: 44255308-44255308 on Build GRCh38
rs1875324 Chr.6: 44255273-44255273 on Build GRCh38
rs199724351 Chr.6: 44255337-44255337 on Build GRCh38
rs756735165 Chr.6: 44255276-44255276 on Build GRCh38
rs749173689 Chr.6: 44255473-44255473 on Build GRCh38
rs755452614 Chr.6: 44255307-44255307 on Build GRCh38
rs774429789 Chr.6: 44255314-44255315 on Build GRCh38
rs147677839 Chr.6: 44255310-44255310 on Build GRCh38
rs145406343 Chr.6: 44255299-44255299 on Build GRCh38
rs1025488976 Chr.6: 44255278-44255278 on Build GRCh38
rs540633770 Chr.6: 44255447-44255447 on Build GRCh38
rs973897805 Chr.6: 44255274-44255274 on Build GRCh38
rs748180992 Chr.6: 44255351-44255352 on Build GRCh38
rs766962448 Chr.6: 44255369-44255369 on Build GRCh38
rs770946830 Chr.6: 44255328-44255328 on Build GRCh38
rs780154680 Chr.6: 44255472-44255472 on Build GRCh38
rs370808683 Chr.6: 44255302-44255302 on Build GRCh38
rs773882742 Chr.6: 44255355-44255355 on Build GRCh38
rs142339274 Chr.6: 44255370-44255370 on Build GRCh38
rs573008456 Chr.6: 44255431-44255431 on Build GRCh38
rs775493883 Chr.6: 44255338-44255338 on Build GRCh38
rs149823072 Chr.6: 44255386-44255386 on Build GRCh38
rs200146778 Chr.6: 44255410-44255410 on Build GRCh38
rs377715053 Chr.6: 44255451-44255451 on Build GRCh38
rs776723576 Chr.6: 44255332-44255332 on Build GRCh38
rs76048013 Chr.6: 44255387-44255387 on Build GRCh38
rs745938537 Chr.6: 44255452-44255452 on Build GRCh38
rs967910881 Chr.6: 44255402-44255402 on Build GRCh38

View Full Product Details