Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196554303-196554538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR53
Entrez Gene ID
348793
Gene Name
WD repeat domain 53
Gene Aliases
-
UniGene
Hs.385865
Gene Chromosome Location
Chr.3: 196554188-196568542 on Build GRCh38
Transcripts
NM_182627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2947381 c.853A>G p.T285A Chr.3: 196554435-196554435 on Build GRCh38
5059616 c.798G>A p.T266T Chr.3: 196554490-196554490 on Build GRCh38
6458929 c.952_953insTTTACC p.K318>IYQ Chr.3: 196554335-196554336 on Build GRCh38
3591873 c.759C>T p.S253S Chr.3: 196554529-196554529 on Build GRCh38
3591872 c.938G>A p.G313D Chr.3: 196554350-196554350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373151770 Chr.3: 196554491-196554491 on Build GRCh38
rs9311 Chr.3: 196554337-196554337 on Build GRCh38
rs149053894 Chr.3: 196554379-196554379 on Build GRCh38
rs148910267 Chr.3: 196554339-196554339 on Build GRCh38
rs995506987 Chr.3: 196554458-196554458 on Build GRCh38
rs377580721 Chr.3: 196554403-196554403 on Build GRCh38
rs752739780 Chr.3: 196554496-196554496 on Build GRCh38
rs752174964 Chr.3: 196554394-196554394 on Build GRCh38
rs775172087 Chr.3: 196554449-196554449 on Build GRCh38
rs746434632 Chr.3: 196554359-196554361 on Build GRCh38
rs766769899 Chr.3: 196554471-196554471 on Build GRCh38
rs1484858 Chr.3: 196554419-196554419 on Build GRCh38
rs566637633 Chr.3: 196554338-196554338 on Build GRCh38
rs778093635 Chr.3: 196554418-196554418 on Build GRCh38
rs369449601 Chr.3: 196554437-196554437 on Build GRCh38
rs748315848 Chr.3: 196554406-196554406 on Build GRCh38
rs144688549 Chr.3: 196554490-196554490 on Build GRCh38
rs779529818 Chr.3: 196554353-196554353 on Build GRCh38
rs760303088 Chr.3: 196554451-196554451 on Build GRCh38
rs763221744 Chr.3: 196554465-196554465 on Build GRCh38
rs758968215 Chr.3: 196554361-196554361 on Build GRCh38
rs868126679 Chr.3: 196554493-196554493 on Build GRCh38
rs145431410 Chr.3: 196554363-196554363 on Build GRCh38
rs754096601 Chr.3: 196554514-196554514 on Build GRCh38
rs756356293 Chr.3: 196554417-196554417 on Build GRCh38
rs745817712 Chr.3: 196554446-196554446 on Build GRCh38
rs766781388 Chr.3: 196554393-196554393 on Build GRCh38
rs887067803 Chr.3: 196554477-196554477 on Build GRCh38
rs61730533 Chr.3: 196554392-196554392 on Build GRCh38
rs749011326 Chr.3: 196554425-196554425 on Build GRCh38
rs767917601 Chr.3: 196554367-196554367 on Build GRCh38
rs771663293 Chr.3: 196554447-196554447 on Build GRCh38
rs1025638509 Chr.3: 196554473-196554473 on Build GRCh38
rs775229474 Chr.3: 196554371-196554371 on Build GRCh38
rs765657934 Chr.3: 196554383-196554383 on Build GRCh38
rs142581925 Chr.3: 196554438-196554438 on Build GRCh38
rs777855201 Chr.3: 196554504-196554504 on Build GRCh38
rs570166175 Chr.3: 196554336-196554337 on Build GRCh38
rs921826131 Chr.3: 196554461-196554461 on Build GRCh38
rs373597954 Chr.3: 196554384-196554384 on Build GRCh38
rs942734465 Chr.3: 196554414-196554414 on Build GRCh38
rs144632277 Chr.3: 196554455-196554455 on Build GRCh38
rs533795093 Chr.3: 196554372-196554372 on Build GRCh38
rs776405241 Chr.3: 196554457-196554457 on Build GRCh38
rs756233337 Chr.3: 196554501-196554501 on Build GRCh38
rs751952027 Chr.3: 196554474-196554474 on Build GRCh38

View Full Product Details