Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44660077-44660330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM53
Entrez Gene ID
79639
Gene Name
transmembrane protein 53
Gene Aliases
NET4
UniGene
Hs.22157
Gene Chromosome Location
Chr.1: 44653247-44674608 on Build GRCh38
Transcripts
NM_001300746.1 NM_001300747.1 NM_001300748.1 NM_024587.3 XM_011542138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5786344 c.171C>T p.I57I Chr.1: 44660186-44660186 on Build GRCh38
5786343 c.171C>T p.I57I Chr.1: 44660186-44660186 on Build GRCh38
3700745 c.120C>G p.L40L Chr.1: 44660237-44660237 on Build GRCh38
5498510 c.142G>T p.D48Y Chr.1: 44660215-44660215 on Build GRCh38
1667520 c.137G>A p.C46Y Chr.1: 44660220-44660220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761438009 Chr.1: 44660124-44660126 on Build GRCh38
rs373159995 Chr.1: 44660166-44660166 on Build GRCh38
rs770974627 Chr.1: 44660133-44660133 on Build GRCh38
rs763837529 Chr.1: 44660160-44660160 on Build GRCh38
rs761618082 Chr.1: 44660163-44660163 on Build GRCh38
rs761708088 Chr.1: 44660290-44660290 on Build GRCh38
rs775313041 Chr.1: 44660142-44660142 on Build GRCh38
rs776361182 Chr.1: 44660283-44660283 on Build GRCh38
rs756469439 Chr.1: 44660217-44660217 on Build GRCh38
rs778046157 Chr.1: 44660218-44660218 on Build GRCh38
rs753172273 Chr.1: 44660186-44660186 on Build GRCh38
rs200638073 Chr.1: 44660139-44660139 on Build GRCh38
rs549222393 Chr.1: 44660162-44660162 on Build GRCh38
rs771939744 Chr.1: 44660140-44660140 on Build GRCh38
rs749510939 Chr.1: 44660132-44660132 on Build GRCh38
rs569249201 Chr.1: 44660184-44660184 on Build GRCh38
rs757541275 Chr.1: 44660226-44660226 on Build GRCh38
rs773017344 Chr.1: 44660308-44660308 on Build GRCh38
rs774471608 Chr.1: 44660137-44660137 on Build GRCh38
rs921519245 Chr.1: 44660202-44660202 on Build GRCh38
rs764920720 Chr.1: 44660292-44660292 on Build GRCh38
rs760480487 Chr.1: 44660151-44660151 on Build GRCh38
rs775468128 Chr.1: 44660279-44660279 on Build GRCh38
rs952582648 Chr.1: 44660109-44660109 on Build GRCh38
rs200411491 Chr.1: 44660281-44660281 on Build GRCh38
rs772077184 Chr.1: 44660259-44660259 on Build GRCh38
rs757991437 Chr.1: 44660178-44660178 on Build GRCh38
rs749890369 Chr.1: 44660175-44660175 on Build GRCh38
rs749499348 Chr.1: 44660224-44660224 on Build GRCh38
rs982899599 Chr.1: 44660114-44660114 on Build GRCh38
rs142353018 Chr.1: 44660257-44660257 on Build GRCh38
rs566868418 Chr.1: 44660106-44660106 on Build GRCh38
rs779122621 Chr.1: 44660234-44660234 on Build GRCh38

View Full Product Details