Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57976295-57976568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEPP
Entrez Gene ID
374739
Gene Name
testis, prostate and placenta expressed
Gene Aliases
-
UniGene
Hs.132976
Gene Chromosome Location
Chr.16: 57976435-57988116 on Build GRCh38
Transcripts
NM_199046.2 NM_199456.2 XM_017023218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479150 c.2T>G p.M1R Chr.16: 57976473-57976473 on Build GRCh38
704019 c.72C>T p.F24F Chr.16: 57976543-57976543 on Build GRCh38
2992554 c.5G>A p.G2D Chr.16: 57976476-57976476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374672165 Chr.16: 57976432-57976432 on Build GRCh38
rs775379442 Chr.16: 57976449-57976449 on Build GRCh38
rs536351256 Chr.16: 57976367-57976367 on Build GRCh38
rs371461112 Chr.16: 57976463-57976463 on Build GRCh38
rs760588932 Chr.16: 57976464-57976464 on Build GRCh38
rs747455015 Chr.16: 57976519-57976519 on Build GRCh38
rs138103950 Chr.16: 57976504-57976504 on Build GRCh38
rs866453780 Chr.16: 57976414-57976414 on Build GRCh38
rs767461521 Chr.16: 57976471-57976471 on Build GRCh38
rs747600125 Chr.16: 57976511-57976511 on Build GRCh38
rs890714271 Chr.16: 57976382-57976382 on Build GRCh38
rs772231188 Chr.16: 57976517-57976517 on Build GRCh38
rs751030983 Chr.16: 57976436-57976436 on Build GRCh38
rs762457626 Chr.16: 57976424-57976424 on Build GRCh38
rs761854153 Chr.16: 57976444-57976444 on Build GRCh38
rs754145698 Chr.16: 57976497-57976497 on Build GRCh38
rs1005425211 Chr.16: 57976389-57976389 on Build GRCh38
rs1056341588 Chr.16: 57976413-57976413 on Build GRCh38
rs750317153 Chr.16: 57976481-57976481 on Build GRCh38
rs762736693 Chr.16: 57976520-57976520 on Build GRCh38
rs755904600 Chr.16: 57976485-57976485 on Build GRCh38
rs761340448 Chr.16: 57976538-57976538 on Build GRCh38
rs35980733 Chr.16: 57976357-57976357 on Build GRCh38
rs951748625 Chr.16: 57976357-57976357 on Build GRCh38
rs763721219 Chr.16: 57976431-57976431 on Build GRCh38
rs996222022 Chr.16: 57976364-57976364 on Build GRCh38
rs913086227 Chr.16: 57976523-57976523 on Build GRCh38
rs939143382 Chr.16: 57976408-57976408 on Build GRCh38
rs375416157 Chr.16: 57976529-57976529 on Build GRCh38
rs750372129 Chr.16: 57976545-57976545 on Build GRCh38
rs766944544 Chr.16: 57976540-57976540 on Build GRCh38
rs920292318 Chr.16: 57976351-57976351 on Build GRCh38
rs185438879 Chr.16: 57976530-57976530 on Build GRCh38
rs779764976 Chr.16: 57976495-57976495 on Build GRCh38
rs900493576 Chr.16: 57976317-57976317 on Build GRCh38
rs375702456 Chr.16: 57976505-57976505 on Build GRCh38
rs149076044 Chr.16: 57976507-57976507 on Build GRCh38
rs1015104399 Chr.16: 57976412-57976412 on Build GRCh38
rs200120066 Chr.16: 57976518-57976518 on Build GRCh38
rs1030993447 Chr.16: 57976368-57976368 on Build GRCh38

View Full Product Details