Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206110288-206110043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
UniGene
Hs.1372
Gene Chromosome Location
Chr.1: 206109849-206117048 on Build GRCh38
Transcripts
NM_000707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444248 c.1220C>A p.P407Q Chr.1: 206110244-206110244 on Build GRCh38
4403160 c.1259A>G p.E420G Chr.1: 206110205-206110205 on Build GRCh38
5520098 c.1260G>A p.E420E Chr.1: 206110204-206110204 on Build GRCh38
7256364 c.1225G>T p.D409Y Chr.1: 206110239-206110239 on Build GRCh38
425139 c.1179C>T p.L393L Chr.1: 206110285-206110285 on Build GRCh38
View More

View Full Product Details