Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53826062-53826319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTO
Entrez Gene ID
79068
Gene Name
fat mass and obesity associated
Gene Aliases
ALKBH9, BMIQ14, GDFD
UniGene
Hs.528833
Gene Chromosome Location
Chr.16: 53703963-54114467 on Build GRCh38
Transcripts
NM_001080432.2 XM_011523313.2 XM_011523314.2 XM_011523315.2 XM_011523316.2 XM_017023654.1 XM_017023655.1 XM_017023656.1 XM_017023657.1 XM_017023658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311241 c.559G>T p.G187* Chr.16: 53826299-53826299 on Build GRCh38
5740651 c.346G>A p.V116I Chr.16: 53826086-53826086 on Build GRCh38
7368920 c.533G>T p.R178M Chr.16: 53826273-53826273 on Build GRCh38
6867208 c.540G>T p.G180G Chr.16: 53826280-53826280 on Build GRCh38
7368919 c.502G>T p.V168L Chr.16: 53826242-53826242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141915969 Chr.16: 53826184-53826184 on Build GRCh38
rs780071553 Chr.16: 53826247-53826247 on Build GRCh38
rs79206939 Chr.16: 53826140-53826140 on Build GRCh38
rs757157849 Chr.16: 53826127-53826127 on Build GRCh38
rs763990762 Chr.16: 53826196-53826196 on Build GRCh38
rs752587241 Chr.16: 53826112-53826112 on Build GRCh38
rs751353294 Chr.16: 53826206-53826206 on Build GRCh38
rs776071679 Chr.16: 53826175-53826175 on Build GRCh38
rs775311728 Chr.16: 53826188-53826188 on Build GRCh38
rs762625200 Chr.16: 53826193-53826193 on Build GRCh38
rs867050060 Chr.16: 53826104-53826104 on Build GRCh38
rs776150628 Chr.16: 53826088-53826088 on Build GRCh38
rs1051308744 Chr.16: 53826157-53826157 on Build GRCh38
rs147561986 Chr.16: 53826168-53826168 on Build GRCh38
rs141423836 Chr.16: 53826135-53826135 on Build GRCh38
rs772503437 Chr.16: 53826290-53826290 on Build GRCh38
rs779309219 Chr.16: 53826267-53826267 on Build GRCh38
rs61743972 Chr.16: 53826285-53826285 on Build GRCh38
rs765812416 Chr.16: 53826085-53826085 on Build GRCh38
rs774633209 Chr.16: 53826160-53826160 on Build GRCh38
rs777330663 Chr.16: 53826097-53826097 on Build GRCh38
rs146941727 Chr.16: 53826154-53826154 on Build GRCh38
rs753133541 Chr.16: 53826189-53826189 on Build GRCh38
rs374337023 Chr.16: 53826117-53826117 on Build GRCh38
rs750522384 Chr.16: 53826241-53826241 on Build GRCh38
rs756242925 Chr.16: 53826245-53826245 on Build GRCh38
rs774450343 Chr.16: 53826098-53826098 on Build GRCh38
rs914343444 Chr.16: 53826119-53826119 on Build GRCh38
rs145884431 Chr.16: 53826227-53826227 on Build GRCh38
rs759168981 Chr.16: 53826089-53826089 on Build GRCh38
rs754065579 Chr.16: 53826249-53826249 on Build GRCh38
rs201792014 Chr.16: 53826114-53826114 on Build GRCh38
rs1031617009 Chr.16: 53826274-53826274 on Build GRCh38
rs898600039 Chr.16: 53826178-53826178 on Build GRCh38
rs745951679 Chr.16: 53826128-53826128 on Build GRCh38
rs867223354 Chr.16: 53826262-53826262 on Build GRCh38
rs547085092 Chr.16: 53826139-53826139 on Build GRCh38
rs201086068 Chr.16: 53826170-53826170 on Build GRCh38
rs371830760 Chr.16: 53826219-53826219 on Build GRCh38
rs367910744 Chr.16: 53826124-53826124 on Build GRCh38
rs762343983 Chr.16: 53826084-53826084 on Build GRCh38
rs755136162 Chr.16: 53826258-53826258 on Build GRCh38
rs182784714 Chr.16: 53826176-53826176 on Build GRCh38
rs751544648 Chr.16: 53826125-53826125 on Build GRCh38

View Full Product Details