Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19624461-19624686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR2
Entrez Gene ID
9170
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
EDG-4, EDG4, LPA-2, LPA2
UniGene
Hs.122575
Gene Chromosome Location
Chr.19: 19623655-19628930 on Build GRCh38
Transcripts
NM_004720.5 XM_011528421.2 XM_017027468.1 XM_017027469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506073 c.769G>A p.G257S Chr.19: 19624543-19624543 on Build GRCh38
1480825 c.781C>T p.L261L Chr.19: 19624531-19624531 on Build GRCh38
3422565 c.750C>T p.F250F Chr.19: 19624562-19624562 on Build GRCh38
3531118 c.795T>C p.G265G Chr.19: 19624517-19624517 on Build GRCh38
993366 c.749T>C p.F250S Chr.19: 19624563-19624563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776164295 Chr.19: 19624609-19624609 on Build GRCh38
rs746002167 Chr.19: 19624534-19624534 on Build GRCh38
rs778235411 Chr.19: 19624513-19624513 on Build GRCh38
rs756150982 Chr.19: 19624545-19624545 on Build GRCh38
rs956037485 Chr.19: 19624645-19624645 on Build GRCh38
rs758974408 Chr.19: 19624591-19624591 on Build GRCh38
rs895874214 Chr.19: 19624580-19624580 on Build GRCh38
rs554252888 Chr.19: 19624616-19624616 on Build GRCh38
rs758694432 Chr.19: 19624515-19624515 on Build GRCh38
rs568372224 Chr.19: 19624633-19624633 on Build GRCh38
rs537365254 Chr.19: 19624661-19624661 on Build GRCh38
rs1031655710 Chr.19: 19624623-19624623 on Build GRCh38
rs775811877 Chr.19: 19624662-19624662 on Build GRCh38
rs781461179 Chr.19: 19624526-19624526 on Build GRCh38
rs751863453 Chr.19: 19624516-19624516 on Build GRCh38
rs778726573 Chr.19: 19624554-19624554 on Build GRCh38
rs34993784 Chr.19: 19624562-19624562 on Build GRCh38
rs986342722 Chr.19: 19624663-19624663 on Build GRCh38
rs200831673 Chr.19: 19624565-19624565 on Build GRCh38
rs763516309 Chr.19: 19624611-19624611 on Build GRCh38
rs868199546 Chr.19: 19624641-19624641 on Build GRCh38
rs758847328 Chr.19: 19624507-19624507 on Build GRCh38
rs368879949 Chr.19: 19624615-19624615 on Build GRCh38
rs773028909 Chr.19: 19624566-19624566 on Build GRCh38
rs753182739 Chr.19: 19624503-19624503 on Build GRCh38
rs1011650430 Chr.19: 19624658-19624658 on Build GRCh38
rs375049332 Chr.19: 19624585-19624585 on Build GRCh38
rs765734644 Chr.19: 19624499-19624499 on Build GRCh38
rs757601036 Chr.19: 19624517-19624517 on Build GRCh38
rs766027293 Chr.19: 19624488-19624488 on Build GRCh38
rs746786744 Chr.19: 19624577-19624577 on Build GRCh38
rs764746400 Chr.19: 19624606-19624606 on Build GRCh38
rs372812887 Chr.19: 19624584-19624584 on Build GRCh38
rs148332097 Chr.19: 19624497-19624497 on Build GRCh38
rs1055980807 Chr.19: 19624567-19624567 on Build GRCh38

View Full Product Details