Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24357613-24357886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
UniGene
Hs.61345
Gene Chromosome Location
Chr.6: 24171755-24383292 on Build GRCh38
Transcripts
NM_001195610.1 NM_016356.4 NM_181337.3

Gene Symbol
KAAG1
Entrez Gene ID
353219
Gene Name
kidney associated antigen 1
Gene Aliases
RU2AS
UniGene
Hs.697888
Gene Chromosome Location
Chr.6: 24356903-24358284 on Build GRCh38
Transcripts
NM_001195610.1 NM_016356.4 NM_181337.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110268 c.247G>A p.E83K Chr.6: 24357886-24357886 on Build GRCh38
6454973 c.105C>T p.A35A Chr.6: 24357744-24357744 on Build GRCh38
3623193 c.108C>T p.I36I Chr.6: 24357643-24357643 on Build GRCh38
741006 c.198G>A p.A66A Chr.6: 24357837-24357837 on Build GRCh38
4877215 c.42C>T p.P14P Chr.6: 24357709-24357709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907477393 Chr.6: 24357814-24357814 on Build GRCh38
rs367903367 Chr.6: 24357791-24357791 on Build GRCh38
rs766573149 Chr.6: 24357727-24357727 on Build GRCh38
rs753069495 Chr.6: 24357749-24357749 on Build GRCh38
rs776505531 Chr.6: 24357833-24357833 on Build GRCh38
rs200664687 Chr.6: 24357806-24357806 on Build GRCh38
rs200585893 Chr.6: 24357839-24357839 on Build GRCh38
rs938050921 Chr.6: 24357658-24357658 on Build GRCh38
rs745436981 Chr.6: 24357785-24357785 on Build GRCh38
rs754074170 Chr.6: 24357739-24357739 on Build GRCh38
rs779618072 Chr.6: 24357657-24357657 on Build GRCh38
rs945462272 Chr.6: 24357704-24357704 on Build GRCh38
rs768455564 Chr.6: 24357697-24357697 on Build GRCh38
rs755386397 Chr.6: 24357652-24357652 on Build GRCh38
rs144637885 Chr.6: 24357698-24357698 on Build GRCh38
rs773192251 Chr.6: 24357815-24357815 on Build GRCh38
rs768259612 Chr.6: 24357803-24357803 on Build GRCh38
rs748676214 Chr.6: 24357666-24357666 on Build GRCh38
rs141716750 Chr.6: 24357821-24357821 on Build GRCh38
rs577487337 Chr.6: 24357694-24357694 on Build GRCh38
rs148029760 Chr.6: 24357706-24357706 on Build GRCh38
rs745333409 Chr.6: 24357683-24357683 on Build GRCh38
rs1054181783 Chr.6: 24357662-24357662 on Build GRCh38
rs199686905 Chr.6: 24357774-24357774 on Build GRCh38
rs201990155 Chr.6: 24357835-24357835 on Build GRCh38
rs757640304 Chr.6: 24357859-24357859 on Build GRCh38
rs773218143 Chr.6: 24357719-24357719 on Build GRCh38
rs903257085 Chr.6: 24357708-24357708 on Build GRCh38
rs370706869 Chr.6: 24357786-24357786 on Build GRCh38
rs538592788 Chr.6: 24357825-24357825 on Build GRCh38
rs780596687 Chr.6: 24357682-24357682 on Build GRCh38
rs774157327 Chr.6: 24357804-24357804 on Build GRCh38
rs1042456481 Chr.6: 24357716-24357716 on Build GRCh38
rs374525825 Chr.6: 24357743-24357743 on Build GRCh38
rs936122381 Chr.6: 24357729-24357729 on Build GRCh38
rs546437721 Chr.6: 24357746-24357746 on Build GRCh38
rs1042640142 Chr.6: 24357700-24357700 on Build GRCh38
rs375996594 Chr.6: 24357644-24357644 on Build GRCh38
rs201096809 Chr.6: 24357855-24357855 on Build GRCh38
rs758995487 Chr.6: 24357769-24357769 on Build GRCh38
rs771948163 Chr.6: 24357811-24357811 on Build GRCh38
rs775204027 Chr.6: 24357690-24357690 on Build GRCh38
rs753161246 Chr.6: 24357836-24357836 on Build GRCh38
rs766491636 Chr.6: 24357828-24357828 on Build GRCh38
rs779647708 Chr.6: 24357864-24357864 on Build GRCh38
rs769381026 Chr.6: 24357688-24357688 on Build GRCh38
rs780304754 Chr.6: 24357636-24357636 on Build GRCh38
rs760773579 Chr.6: 24357724-24357724 on Build GRCh38
rs374324451 Chr.6: 24357787-24357787 on Build GRCh38
rs560979830 Chr.6: 24357842-24357842 on Build GRCh38
rs781394421 Chr.6: 24357809-24357809 on Build GRCh38
rs759867603 Chr.6: 24357834-24357834 on Build GRCh38
rs867724651 Chr.6: 24357638-24357638 on Build GRCh38
rs772638721 Chr.6: 24357678-24357678 on Build GRCh38
rs755190281 Chr.6: 24357745-24357745 on Build GRCh38

View Full Product Details