Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56848643-56848916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKE
Entrez Gene ID
8526
Gene Name
diacylglycerol kinase epsilon
Gene Aliases
AHUS7, DAGK5, DAGK6, DGK, NPHS7
UniGene
Hs.239514
Gene Chromosome Location
Chr.17: 56834099-56869565 on Build GRCh38
Transcripts
NM_003647.2 XM_011525394.2 XM_011525395.2 XM_011525396.2 XM_017025243.1 XM_017025244.1 XM_017025245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848526 c.1027G>A p.D343N Chr.17: 56848834-56848834 on Build GRCh38
6255133 c.960G>T p.L320F Chr.17: 56848767-56848767 on Build GRCh38
3194629 c.966G>A p.W322* Chr.17: 56848773-56848773 on Build GRCh38
4969627 c.967G>A p.G323S Chr.17: 56848774-56848774 on Build GRCh38
3989171 c.1043A>G p.D348G Chr.17: 56848850-56848850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017948303 Chr.17: 56848825-56848825 on Build GRCh38
rs144417124 Chr.17: 56848806-56848806 on Build GRCh38
rs988202789 Chr.17: 56848685-56848685 on Build GRCh38
rs945912552 Chr.17: 56848761-56848761 on Build GRCh38
rs1029933578 Chr.17: 56848798-56848798 on Build GRCh38
rs368125102 Chr.17: 56848817-56848817 on Build GRCh38
rs138924661 Chr.17: 56848773-56848773 on Build GRCh38
rs757336402 Chr.17: 56848853-56848853 on Build GRCh38
rs186080591 Chr.17: 56848862-56848862 on Build GRCh38
rs374898326 Chr.17: 56848785-56848785 on Build GRCh38
rs149433337 Chr.17: 56848805-56848805 on Build GRCh38
rs879255231 Chr.17: 56848694-56848694 on Build GRCh38
rs753872019 Chr.17: 56848852-56848852 on Build GRCh38
rs565388965 Chr.17: 56848813-56848813 on Build GRCh38
rs763712505 Chr.17: 56848854-56848857 on Build GRCh38
rs1038384139 Chr.17: 56848723-56848723 on Build GRCh38
rs147304275 Chr.17: 56848755-56848755 on Build GRCh38
rs777877548 Chr.17: 56848850-56848850 on Build GRCh38
rs772525610 Chr.17: 56848869-56848869 on Build GRCh38
rs749415630 Chr.17: 56848749-56848749 on Build GRCh38
rs372069856 Chr.17: 56848681-56848681 on Build GRCh38
rs756357670 Chr.17: 56848717-56848717 on Build GRCh38
rs762576212 Chr.17: 56848816-56848816 on Build GRCh38
rs767338282 Chr.17: 56848831-56848831 on Build GRCh38
rs566612092 Chr.17: 56848821-56848821 on Build GRCh38
rs777875736 Chr.17: 56848733-56848733 on Build GRCh38
rs138660678 Chr.17: 56848863-56848863 on Build GRCh38
rs312262696 Chr.17: 56848695-56848695 on Build GRCh38
rs756332835 Chr.17: 56848848-56848848 on Build GRCh38
rs746167490 Chr.17: 56848772-56848772 on Build GRCh38
rs754823301 Chr.17: 56848675-56848675 on Build GRCh38
rs774867964 Chr.17: 56848768-56848768 on Build GRCh38
rs752471158 Chr.17: 56848832-56848832 on Build GRCh38
rs312262697 Chr.17: 56848807-56848807 on Build GRCh38
rs748388751 Chr.17: 56848707-56848707 on Build GRCh38
rs200528872 Chr.17: 56848769-56848769 on Build GRCh38
rs371285031 Chr.17: 56848820-56848820 on Build GRCh38

View Full Product Details