Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42896933-42897182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK3
Entrez Gene ID
84376
Gene Name
hook microtubule-tethering protein 3
Gene Aliases
HK3
UniGene
Hs.162852
Gene Chromosome Location
Chr.8: 42896890-43030539 on Build GRCh38
Transcripts
NM_032410.3 XM_011544666.2

Gene Symbol
RNF170
Entrez Gene ID
81790
Gene Name
ring finger protein 170
Gene Aliases
ADSA, SNAX1
UniGene
Hs.491626
Gene Chromosome Location
Chr.8: 42849637-42897288 on Build GRCh38
Transcripts
NM_032410.3 XM_011544666.2

SNP Detail

SNP ID Chromosome Location
rs559730060 Chr.8: 42897018-42897018 on Build GRCh38
rs565805624 Chr.8: 42897017-42897017 on Build GRCh38
rs898851249 Chr.8: 42897080-42897080 on Build GRCh38
rs562561846 Chr.8: 42897126-42897126 on Build GRCh38
rs531446046 Chr.8: 42897131-42897131 on Build GRCh38
rs928043657 Chr.8: 42897064-42897064 on Build GRCh38
rs1016160680 Chr.8: 42897077-42897077 on Build GRCh38
rs913522071 Chr.8: 42896965-42896965 on Build GRCh38
rs1052030870 Chr.8: 42897060-42897060 on Build GRCh38
rs781455902 Chr.8: 42897154-42897154 on Build GRCh38
rs901218127 Chr.8: 42897014-42897014 on Build GRCh38
rs539874332 Chr.8: 42896991-42896991 on Build GRCh38
rs967488336 Chr.8: 42897162-42897162 on Build GRCh38
rs752769156 Chr.8: 42897123-42897124 on Build GRCh38
rs755460322 Chr.8: 42897145-42897145 on Build GRCh38
rs1004791376 Chr.8: 42897068-42897068 on Build GRCh38
rs558074862 Chr.8: 42896988-42896988 on Build GRCh38
rs972531738 Chr.8: 42897159-42897159 on Build GRCh38
rs764915201 Chr.8: 42897100-42897100 on Build GRCh38
rs577893843 Chr.8: 42896989-42896989 on Build GRCh38
rs946282288 Chr.8: 42896983-42896983 on Build GRCh38
rs779657271 Chr.8: 42897118-42897118 on Build GRCh38
rs1039792209 Chr.8: 42897012-42897012 on Build GRCh38
rs556821402 Chr.8: 42897067-42897067 on Build GRCh38
rs769975228 Chr.8: 42897165-42897165 on Build GRCh38
rs866877743 Chr.8: 42897050-42897050 on Build GRCh38
rs748511367 Chr.8: 42897157-42897157 on Build GRCh38
rs951524286 Chr.8: 42897129-42897129 on Build GRCh38
rs372059901 Chr.8: 42897117-42897117 on Build GRCh38
rs367925456 Chr.8: 42897102-42897102 on Build GRCh38
rs960052442 Chr.8: 42897120-42897120 on Build GRCh38

View Full Product Details