Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19879596-19879831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931066 c.2329-2A>G p.? Chr.3: 19879806-19879806 on Build GRCh38
7206466 c.2345G>A p.C782Y Chr.3: 19879788-19879788 on Build GRCh38
86236 c.2333C>T p.A778V Chr.3: 19879800-19879800 on Build GRCh38
1250980 c.2343G>T p.L781F Chr.3: 19879790-19879790 on Build GRCh38
6097586 c.2435A>C p.E812A Chr.3: 19879698-19879698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151124981 Chr.3: 19879656-19879656 on Build GRCh38
rs11917204 Chr.3: 19879657-19879657 on Build GRCh38
rs765943537 Chr.3: 19879773-19879773 on Build GRCh38
rs749338459 Chr.3: 19879685-19879685 on Build GRCh38
rs751749423 Chr.3: 19879711-19879713 on Build GRCh38
rs756597326 Chr.3: 19879668-19879668 on Build GRCh38
rs755106397 Chr.3: 19879725-19879725 on Build GRCh38
rs763005470 Chr.3: 19879769-19879769 on Build GRCh38
rs747735412 Chr.3: 19879635-19879636 on Build GRCh38
rs751081458 Chr.3: 19879783-19879783 on Build GRCh38
rs150316914 Chr.3: 19879661-19879661 on Build GRCh38
rs368362844 Chr.3: 19879669-19879669 on Build GRCh38
rs1038000085 Chr.3: 19879729-19879729 on Build GRCh38
rs527256339 Chr.3: 19879628-19879628 on Build GRCh38
rs772345356 Chr.3: 19879713-19879713 on Build GRCh38
rs774437487 Chr.3: 19879702-19879702 on Build GRCh38
rs915954955 Chr.3: 19879744-19879744 on Build GRCh38
rs941265759 Chr.3: 19879710-19879710 on Build GRCh38
rs144937550 Chr.3: 19879763-19879763 on Build GRCh38
rs111736490 Chr.3: 19879664-19879664 on Build GRCh38
rs754526426 Chr.3: 19879789-19879789 on Build GRCh38
rs775455680 Chr.3: 19879686-19879686 on Build GRCh38
rs908426240 Chr.3: 19879692-19879692 on Build GRCh38
rs373677978 Chr.3: 19879758-19879758 on Build GRCh38
rs746037198 Chr.3: 19879711-19879711 on Build GRCh38
rs748556465 Chr.3: 19879665-19879665 on Build GRCh38
rs767254482 Chr.3: 19879797-19879797 on Build GRCh38
rs189784155 Chr.3: 19879745-19879745 on Build GRCh38
rs761733395 Chr.3: 19879717-19879717 on Build GRCh38
rs765233319 Chr.3: 19879747-19879747 on Build GRCh38
rs371981707 Chr.3: 19879716-19879716 on Build GRCh38
rs758020114 Chr.3: 19879637-19879637 on Build GRCh38
rs766107951 Chr.3: 19879641-19879641 on Build GRCh38

View Full Product Details