Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155023040-155023305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
UniGene
Hs.591491
Gene Chromosome Location
Chr.1: 155018513-155033781 on Build GRCh38
Transcripts
NM_144622.2 XM_011509188.2 XM_011509189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896725 c.2059G>T p.E687* Chr.1: 155023163-155023163 on Build GRCh38
349787 c.1990C>T p.Q664* Chr.1: 155023232-155023232 on Build GRCh38
5377807 c.2020G>A p.D674N Chr.1: 155023202-155023202 on Build GRCh38
4643555 c.2088G>A p.E696E Chr.1: 155023134-155023134 on Build GRCh38
1472703 c.2081C>T p.S694L Chr.1: 155023141-155023141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868046379 Chr.1: 155023174-155023176 on Build GRCh38
rs762323869 Chr.1: 155023137-155023137 on Build GRCh38
rs781261884 Chr.1: 155023163-155023163 on Build GRCh38
rs768064839 Chr.1: 155023209-155023209 on Build GRCh38
rs760281748 Chr.1: 155023071-155023071 on Build GRCh38
rs765746329 Chr.1: 155023219-155023219 on Build GRCh38
rs771671670 Chr.1: 155023076-155023076 on Build GRCh38
rs372646699 Chr.1: 155023089-155023089 on Build GRCh38
rs749192859 Chr.1: 155023170-155023170 on Build GRCh38
rs199629246 Chr.1: 155023084-155023084 on Build GRCh38
rs771759702 Chr.1: 155023187-155023187 on Build GRCh38
rs750141086 Chr.1: 155023284-155023284 on Build GRCh38
rs778601959 Chr.1: 155023179-155023179 on Build GRCh38
rs745720117 Chr.1: 155023259-155023259 on Build GRCh38
rs773764058 Chr.1: 155023226-155023226 on Build GRCh38
rs751751353 Chr.1: 155023159-155023159 on Build GRCh38
rs746686703 Chr.1: 155023265-155023265 on Build GRCh38
rs530964337 Chr.1: 155023263-155023263 on Build GRCh38
rs202210059 Chr.1: 155023145-155023145 on Build GRCh38
rs754305707 Chr.1: 155023125-155023125 on Build GRCh38
rs1055863670 Chr.1: 155023122-155023122 on Build GRCh38
rs763399486 Chr.1: 155023231-155023231 on Build GRCh38
rs1010181393 Chr.1: 155023236-155023236 on Build GRCh38
rs751831385 Chr.1: 155023244-155023244 on Build GRCh38
rs776158967 Chr.1: 155023213-155023213 on Build GRCh38
rs764719772 Chr.1: 155023095-155023095 on Build GRCh38
rs201091113 Chr.1: 155023155-155023155 on Build GRCh38
rs368323169 Chr.1: 155023261-155023261 on Build GRCh38
rs778797991 Chr.1: 155023257-155023257 on Build GRCh38
rs745630100 Chr.1: 155023182-155023182 on Build GRCh38
rs755166177 Chr.1: 155023247-155023247 on Build GRCh38
rs111541777 Chr.1: 155023286-155023286 on Build GRCh38
rs775019621 Chr.1: 155023082-155023082 on Build GRCh38
rs757180720 Chr.1: 155023256-155023256 on Build GRCh38
rs766778547 Chr.1: 155023238-155023238 on Build GRCh38
rs867655193 Chr.1: 155023282-155023282 on Build GRCh38
rs770849267 Chr.1: 155023172-155023172 on Build GRCh38
rs374671082 Chr.1: 155023160-155023160 on Build GRCh38
rs767514129 Chr.1: 155023248-155023248 on Build GRCh38
rs752780038 Chr.1: 155023254-155023254 on Build GRCh38
rs370249543 Chr.1: 155023154-155023154 on Build GRCh38
rs762259367 Chr.1: 155023214-155023214 on Build GRCh38
rs746588465 Chr.1: 155023202-155023202 on Build GRCh38
rs775109444 Chr.1: 155023194-155023194 on Build GRCh38

View Full Product Details