Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38684425-38684668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLO1
Entrez Gene ID
2739
Gene Name
glyoxalase I
Gene Aliases
GLOD1, GLYI, HEL-S-74
UniGene
Hs.268849
Gene Chromosome Location
Chr.6: 38675925-38703176 on Build GRCh38
Transcripts
NM_006708.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1253318 c.216C>T p.F72F Chr.6: 38684466-38684466 on Build GRCh38
4480658 c.243C>T p.I81I Chr.6: 38684439-38684439 on Build GRCh38
3626463 c.190C>T p.P64S Chr.6: 38684492-38684492 on Build GRCh38
6671671 c.236delA p.N79fs*11 Chr.6: 38684446-38684446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141160784 Chr.6: 38684458-38684458 on Build GRCh38
rs762865044 Chr.6: 38684550-38684550 on Build GRCh38
rs11544283 Chr.6: 38684470-38684470 on Build GRCh38
rs965733376 Chr.6: 38684516-38684516 on Build GRCh38
rs748649969 Chr.6: 38684501-38684502 on Build GRCh38
rs772753078 Chr.6: 38684522-38684523 on Build GRCh38
rs770962823 Chr.6: 38684508-38684508 on Build GRCh38
rs775110430 Chr.6: 38684545-38684545 on Build GRCh38
rs778272653 Chr.6: 38684499-38684499 on Build GRCh38
rs530648439 Chr.6: 38684598-38684598 on Build GRCh38
rs779090768 Chr.6: 38684476-38684476 on Build GRCh38
rs910018510 Chr.6: 38684636-38684636 on Build GRCh38
rs755854321 Chr.6: 38684465-38684465 on Build GRCh38
rs550847747 Chr.6: 38684613-38684613 on Build GRCh38
rs76166202 Chr.6: 38684606-38684606 on Build GRCh38
rs113411689 Chr.6: 38684633-38684633 on Build GRCh38
rs759661830 Chr.6: 38684525-38684525 on Build GRCh38
rs748518096 Chr.6: 38684477-38684477 on Build GRCh38
rs543143027 Chr.6: 38684567-38684567 on Build GRCh38
rs923274089 Chr.6: 38684518-38684518 on Build GRCh38
rs984700014 Chr.6: 38684620-38684620 on Build GRCh38
rs376656094 Chr.6: 38684532-38684532 on Build GRCh38
rs570771772 Chr.6: 38684618-38684618 on Build GRCh38
rs772527200 Chr.6: 38684498-38684498 on Build GRCh38
rs747563298 Chr.6: 38684501-38684501 on Build GRCh38
rs529346728 Chr.6: 38684511-38684511 on Build GRCh38
rs776834145 Chr.6: 38684524-38684524 on Build GRCh38

View Full Product Details