Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34866106-34866355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP3
Entrez Gene ID
58512
Gene Name
DLG associated protein 3
Gene Aliases
DAP3, SAPAP3
UniGene
Hs.733945
Gene Chromosome Location
Chr.1: 34862120-34929585 on Build GRCh38
Transcripts
NM_001080418.2 XM_011541879.2 XM_011541880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182581 c.2879C>T p.S960L Chr.1: 34866144-34866144 on Build GRCh38
4031753 c.2893G>A p.A965T Chr.1: 34866130-34866130 on Build GRCh38
6644973 c.2882C>T p.A961V Chr.1: 34866141-34866141 on Build GRCh38
6392018 c.2852A>T p.K951M Chr.1: 34866171-34866171 on Build GRCh38
6379869 c.2813G>A p.R938Q Chr.1: 34866210-34866210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763403379 Chr.1: 34866262-34866262 on Build GRCh38
rs772306893 Chr.1: 34866221-34866221 on Build GRCh38
rs1025962316 Chr.1: 34866257-34866257 on Build GRCh38
rs762256166 Chr.1: 34866252-34866252 on Build GRCh38
rs908946337 Chr.1: 34866291-34866291 on Build GRCh38
rs780230575 Chr.1: 34866223-34866223 on Build GRCh38
rs765923493 Chr.1: 34866279-34866279 on Build GRCh38
rs776921512 Chr.1: 34866243-34866243 on Build GRCh38
rs751040025 Chr.1: 34866281-34866281 on Build GRCh38
rs777777768 Chr.1: 34866215-34866215 on Build GRCh38
rs752531018 Chr.1: 34866185-34866185 on Build GRCh38
rs990310573 Chr.1: 34866320-34866320 on Build GRCh38
rs754603347 Chr.1: 34866158-34866158 on Build GRCh38
rs775105549 Chr.1: 34866296-34866298 on Build GRCh38
rs752231831 Chr.1: 34866308-34866308 on Build GRCh38
rs539291311 Chr.1: 34866187-34866187 on Build GRCh38
rs762457264 Chr.1: 34866143-34866143 on Build GRCh38
rs769989521 Chr.1: 34866254-34866254 on Build GRCh38
rs767184922 Chr.1: 34866307-34866307 on Build GRCh38
rs866547644 Chr.1: 34866296-34866296 on Build GRCh38
rs780890665 Chr.1: 34866181-34866181 on Build GRCh38
rs994167462 Chr.1: 34866236-34866236 on Build GRCh38
rs749112086 Chr.1: 34866218-34866218 on Build GRCh38
rs949627203 Chr.1: 34866314-34866314 on Build GRCh38
rs777387655 Chr.1: 34866316-34866316 on Build GRCh38
rs867317403 Chr.1: 34866258-34866258 on Build GRCh38
rs768964682 Chr.1: 34866242-34866242 on Build GRCh38
rs765837114 Chr.1: 34866154-34866154 on Build GRCh38
rs533602443 Chr.1: 34866314-34866314 on Build GRCh38
rs566251521 Chr.1: 34866289-34866289 on Build GRCh38
rs1056555905 Chr.1: 34866288-34866288 on Build GRCh38
rs773803655 Chr.1: 34866261-34866261 on Build GRCh38
rs759540010 Chr.1: 34866278-34866278 on Build GRCh38
rs913528214 Chr.1: 34866325-34866325 on Build GRCh38
rs897739081 Chr.1: 34866153-34866153 on Build GRCh38
rs747280130 Chr.1: 34866226-34866226 on Build GRCh38
rs557496191 Chr.1: 34866284-34866284 on Build GRCh38
rs1001638853 Chr.1: 34866309-34866309 on Build GRCh38
rs937816972 Chr.1: 34866292-34866292 on Build GRCh38
rs1024309598 Chr.1: 34866237-34866237 on Build GRCh38
rs771547387 Chr.1: 34866290-34866292 on Build GRCh38
rs753847027 Chr.1: 34866335-34866335 on Build GRCh38

View Full Product Details