Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149723671-149723944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRBA1
Entrez Gene ID
84626
Gene Name
KRAB-A domain containing 1
Gene Aliases
-
UniGene
Hs.299560
Gene Chromosome Location
Chr.7: 149714945-149734575 on Build GRCh38
Transcripts
NM_001290187.1 NM_032534.3 XM_011516636.2 XM_011516637.2 XM_011516638.2 XM_011516639.2 XM_011516640.2 XM_011516641.2 XM_011516642.2 XM_011516643.2

SNP Detail

SNP ID Chromosome Location
rs774241835 Chr.7: 149723742-149723742 on Build GRCh38
rs757707289 Chr.7: 149723854-149723854 on Build GRCh38
rs768015585 Chr.7: 149723897-149723897 on Build GRCh38
rs748854457 Chr.7: 149723896-149723896 on Build GRCh38
rs560584214 Chr.7: 149723902-149723902 on Build GRCh38
rs371071905 Chr.7: 149723894-149723894 on Build GRCh38
rs547173880 Chr.7: 149723852-149723852 on Build GRCh38
rs770918859 Chr.7: 149723913-149723913 on Build GRCh38
rs369939735 Chr.7: 149723747-149723747 on Build GRCh38
rs76019230 Chr.7: 149723904-149723904 on Build GRCh38
rs889537639 Chr.7: 149723880-149723880 on Build GRCh38
rs139849988 Chr.7: 149723920-149723920 on Build GRCh38
rs765306211 Chr.7: 149723780-149723780 on Build GRCh38
rs368713703 Chr.7: 149723903-149723903 on Build GRCh38
rs376659891 Chr.7: 149723744-149723744 on Build GRCh38
rs749937432 Chr.7: 149723728-149723729 on Build GRCh38
rs372393153 Chr.7: 149723792-149723792 on Build GRCh38
rs751764004 Chr.7: 149723850-149723850 on Build GRCh38
rs776389730 Chr.7: 149723918-149723918 on Build GRCh38
rs748716706 Chr.7: 149723813-149723813 on Build GRCh38
rs771611930 Chr.7: 149723721-149723721 on Build GRCh38
rs775134331 Chr.7: 149723811-149723811 on Build GRCh38
rs750814351 Chr.7: 149723873-149723873 on Build GRCh38
rs375688146 Chr.7: 149723807-149723807 on Build GRCh38
rs777228436 Chr.7: 149723726-149723726 on Build GRCh38
rs928560827 Chr.7: 149723755-149723755 on Build GRCh38
rs34685827 Chr.7: 149723887-149723888 on Build GRCh38
rs11462346 Chr.7: 149723729-149723730 on Build GRCh38
rs939979330 Chr.7: 149723770-149723770 on Build GRCh38
rs767467991 Chr.7: 149723746-149723746 on Build GRCh38
rs752712153 Chr.7: 149723782-149723782 on Build GRCh38
rs777832304 Chr.7: 149723787-149723787 on Build GRCh38
rs200547612 Chr.7: 149723729-149723729 on Build GRCh38
rs370272934 Chr.7: 149723821-149723821 on Build GRCh38
rs990871668 Chr.7: 149723716-149723716 on Build GRCh38
rs397890289 Chr.7: 149723731-149723732 on Build GRCh38
rs761632549 Chr.7: 149723832-149723832 on Build GRCh38
rs745561227 Chr.7: 149723801-149723801 on Build GRCh38
rs775018272 Chr.7: 149723734-149723734 on Build GRCh38
rs376059171 Chr.7: 149723868-149723868 on Build GRCh38
rs773149774 Chr.7: 149723843-149723843 on Build GRCh38
rs747300403 Chr.7: 149723714-149723714 on Build GRCh38
rs776460640 Chr.7: 149723767-149723767 on Build GRCh38
rs760669890 Chr.7: 149723844-149723844 on Build GRCh38
rs374326942 Chr.7: 149723900-149723900 on Build GRCh38
rs778265263 Chr.7: 149723702-149723702 on Build GRCh38
rs755058019 Chr.7: 149723768-149723768 on Build GRCh38
rs758684405 Chr.7: 149723784-149723784 on Build GRCh38
rs200134914 Chr.7: 149723847-149723847 on Build GRCh38
rs756686737 Chr.7: 149723878-149723878 on Build GRCh38
rs767235088 Chr.7: 149723840-149723840 on Build GRCh38
rs200100018 Chr.7: 149723727-149723727 on Build GRCh38
rs766194216 Chr.7: 149723845-149723845 on Build GRCh38
rs764407814 Chr.7: 149723848-149723848 on Build GRCh38
rs757655966 Chr.7: 149723800-149723800 on Build GRCh38
rs958093482 Chr.7: 149723693-149723693 on Build GRCh38
rs1052242602 Chr.7: 149723921-149723921 on Build GRCh38

View Full Product Details