Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24022336-24022607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD2B
Entrez Gene ID
388931
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
-
UniGene
Hs.407482
Gene Chromosome Location
Chr.2: 24009022-24026775 on Build GRCh38
Transcripts
NM_001080473.1 XM_005264301.4 XM_017004063.1 XM_017004064.1 XM_017004065.1 XM_017004066.1 XM_017004067.1 XM_017004068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3372717 c.927A>G p.T309T Chr.2: 24022465-24022465 on Build GRCh38
5106490 c.894+79_978+37del505 p.? Chr.2: 24022049-24022553 on Build GRCh38
2910707 c.969A>C p.L323L Chr.2: 24022507-24022507 on Build GRCh38
1406792 c.953T>C p.V318A Chr.2: 24022491-24022491 on Build GRCh38
720836 c.949C>T p.H317Y Chr.2: 24022487-24022487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773613843 Chr.2: 24022555-24022555 on Build GRCh38
rs755242058 Chr.2: 24022520-24022520 on Build GRCh38
rs780121003 Chr.2: 24022438-24022438 on Build GRCh38
rs963846209 Chr.2: 24022559-24022559 on Build GRCh38
rs757203000 Chr.2: 24022506-24022506 on Build GRCh38
rs1050044962 Chr.2: 24022571-24022571 on Build GRCh38
rs1021467823 Chr.2: 24022443-24022443 on Build GRCh38
rs146632569 Chr.2: 24022543-24022543 on Build GRCh38
rs773596912 Chr.2: 24022467-24022467 on Build GRCh38
rs747286855 Chr.2: 24022452-24022452 on Build GRCh38
rs759305073 Chr.2: 24022490-24022490 on Build GRCh38
rs749828864 Chr.2: 24022557-24022557 on Build GRCh38
rs754665740 Chr.2: 24022398-24022398 on Build GRCh38
rs144012352 Chr.2: 24022410-24022410 on Build GRCh38
rs775011370 Chr.2: 24022519-24022519 on Build GRCh38
rs777597523 Chr.2: 24022413-24022413 on Build GRCh38
rs940143976 Chr.2: 24022531-24022531 on Build GRCh38
rs890836433 Chr.2: 24022362-24022362 on Build GRCh38
rs530457898 Chr.2: 24022406-24022406 on Build GRCh38
rs759329509 Chr.2: 24022497-24022497 on Build GRCh38
rs758409263 Chr.2: 24022434-24022434 on Build GRCh38
rs376661104 Chr.2: 24022423-24022423 on Build GRCh38
rs781414682 Chr.2: 24022538-24022538 on Build GRCh38
rs970384931 Chr.2: 24022459-24022459 on Build GRCh38
rs767490645 Chr.2: 24022400-24022400 on Build GRCh38
rs763877773 Chr.2: 24022501-24022501 on Build GRCh38
rs755888766 Chr.2: 24022499-24022499 on Build GRCh38
rs771879358 Chr.2: 24022529-24022529 on Build GRCh38
rs752511553 Chr.2: 24022409-24022409 on Build GRCh38
rs377058845 Chr.2: 24022502-24022502 on Build GRCh38
rs748489541 Chr.2: 24022464-24022464 on Build GRCh38
rs968733817 Chr.2: 24022540-24022540 on Build GRCh38
rs745690268 Chr.2: 24022482-24022484 on Build GRCh38
rs377638611 Chr.2: 24022465-24022465 on Build GRCh38
rs374005294 Chr.2: 24022435-24022435 on Build GRCh38
rs780173246 Chr.2: 24022507-24022507 on Build GRCh38
rs779832165 Chr.2: 24022584-24022584 on Build GRCh38
rs768860750 Chr.2: 24022457-24022457 on Build GRCh38
rs777018908 Chr.2: 24022460-24022460 on Build GRCh38
rs749206666 Chr.2: 24022422-24022422 on Build GRCh38
rs1008386264 Chr.2: 24022386-24022386 on Build GRCh38
rs747058281 Chr.2: 24022516-24022516 on Build GRCh38
rs976520041 Chr.2: 24022567-24022567 on Build GRCh38
rs770068070 Chr.2: 24022548-24022548 on Build GRCh38
rs374381466 Chr.2: 24022489-24022489 on Build GRCh38
rs371432365 Chr.2: 24022483-24022483 on Build GRCh38

View Full Product Details