Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49430950-49431207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A7
Entrez Gene ID
57030
Gene Name
solute carrier family 17 member 7
Gene Aliases
BNPI, VGLUT1
UniGene
Hs.375616
Gene Chromosome Location
Chr.19: 49429398-49441551 on Build GRCh38
Transcripts
NM_020309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6525177 c.1365C>T p.I455I Chr.19: 49431039-49431039 on Build GRCh38
1729207 c.1286T>A p.I429K Chr.19: 49431118-49431118 on Build GRCh38
7207443 c.1339C>T p.L447L Chr.19: 49431065-49431065 on Build GRCh38
3404454 c.1292C>T p.P431L Chr.19: 49431112-49431112 on Build GRCh38
6151965 c.1329C>T p.G443G Chr.19: 49431075-49431075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777496101 Chr.19: 49431154-49431155 on Build GRCh38
rs750805407 Chr.19: 49431142-49431142 on Build GRCh38
rs749233541 Chr.19: 49431177-49431177 on Build GRCh38
rs375622644 Chr.19: 49430989-49430989 on Build GRCh38
rs767792001 Chr.19: 49431072-49431072 on Build GRCh38
rs779470522 Chr.19: 49430990-49430990 on Build GRCh38
rs765801106 Chr.19: 49431135-49431135 on Build GRCh38
rs748129512 Chr.19: 49431038-49431038 on Build GRCh38
rs368231298 Chr.19: 49430997-49430997 on Build GRCh38
rs546901660 Chr.19: 49431057-49431057 on Build GRCh38
rs760947355 Chr.19: 49431096-49431096 on Build GRCh38
rs746518221 Chr.19: 49431174-49431174 on Build GRCh38
rs980879501 Chr.19: 49431029-49431029 on Build GRCh38
rs371139022 Chr.19: 49430986-49430986 on Build GRCh38
rs375230026 Chr.19: 49431161-49431161 on Build GRCh38
rs780699580 Chr.19: 49431165-49431165 on Build GRCh38
rs776117731 Chr.19: 49431033-49431033 on Build GRCh38
rs762433312 Chr.19: 49431129-49431129 on Build GRCh38
rs775630264 Chr.19: 49431092-49431092 on Build GRCh38
rs753945868 Chr.19: 49431111-49431111 on Build GRCh38
rs528346148 Chr.19: 49431021-49431021 on Build GRCh38
rs758102698 Chr.19: 49431005-49431005 on Build GRCh38
rs769666998 Chr.19: 49431047-49431047 on Build GRCh38
rs758635682 Chr.19: 49431148-49431148 on Build GRCh38
rs780455901 Chr.19: 49431160-49431160 on Build GRCh38
rs532058755 Chr.19: 49431110-49431110 on Build GRCh38
rs971797832 Chr.19: 49431078-49431078 on Build GRCh38
rs987464218 Chr.19: 49430977-49430977 on Build GRCh38
rs777627352 Chr.19: 49431176-49431176 on Build GRCh38
rs532385556 Chr.19: 49431188-49431188 on Build GRCh38
rs769724199 Chr.19: 49431172-49431172 on Build GRCh38
rs17855709 Chr.19: 49431112-49431112 on Build GRCh38
rs368670946 Chr.19: 49431187-49431187 on Build GRCh38
rs60408689 Chr.19: 49431006-49431006 on Build GRCh38
rs1035279684 Chr.19: 49431030-49431030 on Build GRCh38
rs746545092 Chr.19: 49431010-49431010 on Build GRCh38
rs747612607 Chr.19: 49431168-49431168 on Build GRCh38
rs919699130 Chr.19: 49431048-49431048 on Build GRCh38
rs138689882 Chr.19: 49431109-49431109 on Build GRCh38
rs143015348 Chr.19: 49431051-49431051 on Build GRCh38

View Full Product Details