Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158663214-158663471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
UniGene
Hs.436977
Gene Chromosome Location
Chr.6: 158644881-158766298 on Build GRCh38
Transcripts
NM_001009991.3 NM_001242384.1 NM_001242394.1 NM_001242395.1 NM_001318745.1 XM_005267215.4 XM_005267218.4 XM_006715605.3 XM_006715606.3 XM_006715611.3 XM_011536254.2 XM_017011495.1 XM_017011496.1 XM_017011497.1 XM_017011498.1 XM_017011499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582667 c.70C>T p.R24* Chr.6: 158663338-158663338 on Build GRCh38
6848868 c.55C>T p.L19F Chr.6: 158663323-158663323 on Build GRCh38
6514594 c.46G>A p.E16K Chr.6: 158663314-158663314 on Build GRCh38
3829558 c.46G>C p.E16Q Chr.6: 158663314-158663314 on Build GRCh38
3829559 c.46G>C p.E16Q Chr.6: 158663314-158663314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769350781 Chr.6: 158663354-158663354 on Build GRCh38
rs996423873 Chr.6: 158663401-158663401 on Build GRCh38
rs899026933 Chr.6: 158663386-158663386 on Build GRCh38
rs764284992 Chr.6: 158663269-158663269 on Build GRCh38
rs754352151 Chr.6: 158663411-158663411 on Build GRCh38
rs951567314 Chr.6: 158663361-158663362 on Build GRCh38
rs770410741 Chr.6: 158663302-158663302 on Build GRCh38
rs527438917 Chr.6: 158663364-158663364 on Build GRCh38
rs751624665 Chr.6: 158663275-158663275 on Build GRCh38
rs751238338 Chr.6: 158663425-158663425 on Build GRCh38
rs750970788 Chr.6: 158663336-158663336 on Build GRCh38
rs541677337 Chr.6: 158663344-158663344 on Build GRCh38
rs560281807 Chr.6: 158663349-158663349 on Build GRCh38
rs2291384 Chr.6: 158663253-158663253 on Build GRCh38
rs936908567 Chr.6: 158663270-158663270 on Build GRCh38
rs762086061 Chr.6: 158663394-158663394 on Build GRCh38
rs757410330 Chr.6: 158663278-158663278 on Build GRCh38
rs574758010 Chr.6: 158663314-158663314 on Build GRCh38
rs139996805 Chr.6: 158663311-158663311 on Build GRCh38
rs750944778 Chr.6: 158663282-158663282 on Build GRCh38
rs371220640 Chr.6: 158663339-158663339 on Build GRCh38
rs772658315 Chr.6: 158663235-158663235 on Build GRCh38
rs769555106 Chr.6: 158663360-158663360 on Build GRCh38
rs762033124 Chr.6: 158663328-158663328 on Build GRCh38
rs777266435 Chr.6: 158663291-158663291 on Build GRCh38
rs747856881 Chr.6: 158663286-158663286 on Build GRCh38
rs770463652 Chr.6: 158663348-158663348 on Build GRCh38
rs545871542 Chr.6: 158663407-158663407 on Build GRCh38
rs767672844 Chr.6: 158663332-158663332 on Build GRCh38
rs746586449 Chr.6: 158663292-158663292 on Build GRCh38
rs144534535 Chr.6: 158663283-158663283 on Build GRCh38
rs201251694 Chr.6: 158663288-158663288 on Build GRCh38
rs767468785 Chr.6: 158663284-158663284 on Build GRCh38
rs746637702 Chr.6: 158663346-158663346 on Build GRCh38
rs555111308 Chr.6: 158663250-158663250 on Build GRCh38
rs978662760 Chr.6: 158663265-158663265 on Build GRCh38
rs762716551 Chr.6: 158663377-158663377 on Build GRCh38
rs376260554 Chr.6: 158663378-158663378 on Build GRCh38
rs534279054 Chr.6: 158663259-158663259 on Build GRCh38
rs754080810 Chr.6: 158663341-158663341 on Build GRCh38
rs748251374 Chr.6: 158663366-158663367 on Build GRCh38
rs1028890300 Chr.6: 158663444-158663444 on Build GRCh38
rs749768882 Chr.6: 158663286-158663287 on Build GRCh38
rs774555163 Chr.6: 158663357-158663358 on Build GRCh38
rs765551329 Chr.6: 158663416-158663416 on Build GRCh38
rs764479125 Chr.6: 158663316-158663316 on Build GRCh38
rs374316566 Chr.6: 158663260-158663260 on Build GRCh38
rs200370553 Chr.6: 158663313-158663313 on Build GRCh38
rs751820898 Chr.6: 158663319-158663319 on Build GRCh38
rs766858408 Chr.6: 158663400-158663400 on Build GRCh38
rs949130308 Chr.6: 158663446-158663446 on Build GRCh38
rs201715127 Chr.6: 158663338-158663338 on Build GRCh38
rs377730182 Chr.6: 158663375-158663375 on Build GRCh38
rs745833675 Chr.6: 158663358-158663358 on Build GRCh38
rs755381449 Chr.6: 158663414-158663414 on Build GRCh38
rs756848096 Chr.6: 158663429-158663429 on Build GRCh38
rs916851143 Chr.6: 158663294-158663294 on Build GRCh38
rs577302182 Chr.6: 158663304-158663304 on Build GRCh38
rs780328906 Chr.6: 158663267-158663268 on Build GRCh38
rs781779174 Chr.6: 158663280-158663280 on Build GRCh38
rs773105838 Chr.6: 158663395-158663395 on Build GRCh38
rs752998958 Chr.6: 158663266-158663266 on Build GRCh38

View Full Product Details