Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100851136-100851397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOX1
Entrez Gene ID
27035
Gene Name
NADPH oxidase 1
Gene Aliases
GP91-2, MOX1, NOH-1, NOH1
UniGene
Hs.592227
Gene Chromosome Location
Chr.X: 100843324-100874345 on Build GRCh38
Transcripts
NM_001271815.1 NM_007052.4 NM_013955.2 XM_017029407.1

SNP Detail

SNP ID Chromosome Location
rs148147755 Chr.X: 100851291-100851291 on Build GRCh38
rs760026368 Chr.X: 100851196-100851197 on Build GRCh38
rs376777923 Chr.X: 100851246-100851246 on Build GRCh38
rs373159412 Chr.X: 100851183-100851183 on Build GRCh38
rs757706919 Chr.X: 100851191-100851191 on Build GRCh38
rs758629194 Chr.X: 100851356-100851356 on Build GRCh38
rs763073506 Chr.X: 100851304-100851304 on Build GRCh38
rs781545560 Chr.X: 100851207-100851207 on Build GRCh38
rs914664208 Chr.X: 100851241-100851241 on Build GRCh38
rs1031136457 Chr.X: 100851242-100851242 on Build GRCh38
rs772834730 Chr.X: 100851261-100851261 on Build GRCh38
rs1001043781 Chr.X: 100851224-100851224 on Build GRCh38
rs143748762 Chr.X: 100851287-100851287 on Build GRCh38
rs369091802 Chr.X: 100851332-100851332 on Build GRCh38
rs143127702 Chr.X: 100851270-100851270 on Build GRCh38
rs904098486 Chr.X: 100851160-100851160 on Build GRCh38
rs754612284 Chr.X: 100851217-100851217 on Build GRCh38
rs139091669 Chr.X: 100851238-100851238 on Build GRCh38
rs746955554 Chr.X: 100851268-100851268 on Build GRCh38
rs778984604 Chr.X: 100851252-100851252 on Build GRCh38
rs746193953 Chr.X: 100851211-100851211 on Build GRCh38
rs778673989 Chr.X: 100851222-100851222 on Build GRCh38
rs927234631 Chr.X: 100851363-100851363 on Build GRCh38
rs752120286 Chr.X: 100851184-100851184 on Build GRCh38
rs576448500 Chr.X: 100851303-100851303 on Build GRCh38
rs764250572 Chr.X: 100851328-100851328 on Build GRCh38

View Full Product Details