Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1171709-1171940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPON2
Entrez Gene ID
10417
Gene Name
spondin 2
Gene Aliases
DIL-1, DIL1, M-SPONDIN, MINDIN
UniGene
Hs.302963
Gene Chromosome Location
Chr.4: 1166932-1208962 on Build GRCh38
Transcripts
NM_001128325.2 NM_001199021.1 NM_012445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6511947 c.138C>T p.T46T Chr.4: 1171934-1171934 on Build GRCh38
301466 c.220_220+1GG>TT p.? Chr.4: 1171851-1171852 on Build GRCh38
1050309 c.212C>T p.S71L Chr.4: 1171860-1171860 on Build GRCh38
6438357 c.162_163GG>AA p.A55T Chr.4: 1171909-1171910 on Build GRCh38
4121534 c.201G>A p.A67A Chr.4: 1171871-1171871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527258181 Chr.4: 1171745-1171745 on Build GRCh38
rs965412474 Chr.4: 1171888-1171888 on Build GRCh38
rs928735611 Chr.4: 1171837-1171837 on Build GRCh38
rs376180391 Chr.4: 1171910-1171910 on Build GRCh38
rs371203962 Chr.4: 1171872-1171872 on Build GRCh38
rs1007691424 Chr.4: 1171755-1171755 on Build GRCh38
rs895044347 Chr.4: 1171794-1171794 on Build GRCh38
rs748354801 Chr.4: 1171876-1171876 on Build GRCh38
rs1017278255 Chr.4: 1171756-1171756 on Build GRCh38
rs951792251 Chr.4: 1171749-1171749 on Build GRCh38
rs199893739 Chr.4: 1171810-1171810 on Build GRCh38
rs767171681 Chr.4: 1171839-1171839 on Build GRCh38
rs769827720 Chr.4: 1171812-1171812 on Build GRCh38
rs769926787 Chr.4: 1171871-1171871 on Build GRCh38
rs775922429 Chr.4: 1171874-1171875 on Build GRCh38
rs761936028 Chr.4: 1171811-1171811 on Build GRCh38
rs766117678 Chr.4: 1171830-1171830 on Build GRCh38
rs577165534 Chr.4: 1171918-1171918 on Build GRCh38
rs750892979 Chr.4: 1171846-1171846 on Build GRCh38
rs573292587 Chr.4: 1171874-1171874 on Build GRCh38
rs559042658 Chr.4: 1171897-1171897 on Build GRCh38
rs897432543 Chr.4: 1171873-1171873 on Build GRCh38
rs562953830 Chr.4: 1171903-1171903 on Build GRCh38
rs529981694 Chr.4: 1171851-1171851 on Build GRCh38
rs1011684759 Chr.4: 1171831-1171831 on Build GRCh38
rs760773700 Chr.4: 1171887-1171888 on Build GRCh38
rs753290725 Chr.4: 1171912-1171912 on Build GRCh38
rs2242278 Chr.4: 1171728-1171728 on Build GRCh38
rs772706163 Chr.4: 1171873-1171885 on Build GRCh38
rs566316968 Chr.4: 1171741-1171741 on Build GRCh38
rs762091208 Chr.4: 1171746-1171746 on Build GRCh38
rs571597094 Chr.4: 1171795-1171795 on Build GRCh38
rs975766100 Chr.4: 1171768-1171768 on Build GRCh38
rs781469354 Chr.4: 1171875-1171875 on Build GRCh38
rs2242279 Chr.4: 1171799-1171799 on Build GRCh38
rs751389880 Chr.4: 1171835-1171835 on Build GRCh38
rs1032165821 Chr.4: 1171824-1171824 on Build GRCh38
rs955939652 Chr.4: 1171796-1171796 on Build GRCh38
rs377667252 Chr.4: 1171838-1171838 on Build GRCh38
rs773191454 Chr.4: 1171877-1171877 on Build GRCh38
rs758917086 Chr.4: 1171914-1171914 on Build GRCh38
rs747390530 Chr.4: 1171863-1171863 on Build GRCh38
rs150653050 Chr.4: 1171853-1171853 on Build GRCh38
rs755646798 Chr.4: 1171841-1171841 on Build GRCh38
rs1003132059 Chr.4: 1171740-1171740 on Build GRCh38
rs200210532 Chr.4: 1171814-1171814 on Build GRCh38
rs867296831 Chr.4: 1171762-1171762 on Build GRCh38
rs868378454 Chr.4: 1171743-1171743 on Build GRCh38
rs144576850 Chr.4: 1171892-1171892 on Build GRCh38
rs914379955 Chr.4: 1171805-1171805 on Build GRCh38
rs370307267 Chr.4: 1171748-1171748 on Build GRCh38
rs972854047 Chr.4: 1171730-1171730 on Build GRCh38
rs1048286491 Chr.4: 1171792-1171792 on Build GRCh38
rs201763485 Chr.4: 1171847-1171847 on Build GRCh38
rs147457530 Chr.4: 1171904-1171904 on Build GRCh38

View Full Product Details