Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113592882-113593155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIDT1
Entrez Gene ID
54847
Gene Name
SID1 transmembrane family member 1
Gene Aliases
SID-1, SID1
UniGene
Hs.591291
Gene Chromosome Location
Chr.3: 113531774-113629579 on Build GRCh38
Transcripts
NM_001308350.1 NM_001322294.1 NM_001322295.1 NM_001322296.1 NM_001322297.1 NM_001322298.1 NM_001322299.1 NM_001322300.1 NM_017699.2 NR_136280.1 NR_136281.1 NR_136282.1 NR_136283.1 NR_136284.1 NR_136285.1 XM_011512939.2 XM_011512941.2 XM_017006664.1 XM_017006665.1 XM_017006666.1 XM_017006667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6477507 c.1002-2A>T p.? Chr.3: 113593003-113593003 on Build GRCh38
6477508 c.1002-2A>T p.? Chr.3: 113593003-113593003 on Build GRCh38
131306 c.1030T>A p.F344I Chr.3: 113593033-113593033 on Build GRCh38
5023484 c.1045+2T>C p.? Chr.3: 113593050-113593050 on Build GRCh38
5023485 c.1045+2T>C p.? Chr.3: 113593050-113593050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768185278 Chr.3: 113593008-113593008 on Build GRCh38
rs888348643 Chr.3: 113593104-113593104 on Build GRCh38
rs1011711745 Chr.3: 113592904-113592904 on Build GRCh38
rs774836637 Chr.3: 113593005-113593005 on Build GRCh38
rs775284679 Chr.3: 113592998-113592998 on Build GRCh38
rs564297351 Chr.3: 113592915-113592915 on Build GRCh38
rs78339526 Chr.3: 113592905-113592905 on Build GRCh38
rs1030876427 Chr.3: 113592909-113592909 on Build GRCh38
rs757637840 Chr.3: 113592974-113592974 on Build GRCh38
rs761614142 Chr.3: 113593013-113593013 on Build GRCh38
rs751764063 Chr.3: 113593056-113593056 on Build GRCh38
rs202200078 Chr.3: 113593047-113593047 on Build GRCh38
rs774194071 Chr.3: 113593011-113593011 on Build GRCh38
rs781765595 Chr.3: 113592977-113592977 on Build GRCh38
rs756545567 Chr.3: 113593086-113593086 on Build GRCh38
rs778323156 Chr.3: 113592954-113592954 on Build GRCh38
rs560570434 Chr.3: 113592907-113592907 on Build GRCh38
rs752689521 Chr.3: 113593043-113593043 on Build GRCh38
rs367921267 Chr.3: 113592978-113592978 on Build GRCh38
rs956283266 Chr.3: 113592924-113592924 on Build GRCh38
rs779678080 Chr.3: 113593088-113593088 on Build GRCh38
rs760095152 Chr.3: 113592938-113592938 on Build GRCh38
rs147404509 Chr.3: 113593028-113593028 on Build GRCh38
rs989096073 Chr.3: 113592973-113592973 on Build GRCh38
rs927419701 Chr.3: 113593015-113593015 on Build GRCh38
rs747374300 Chr.3: 113592963-113592963 on Build GRCh38
rs765408617 Chr.3: 113593038-113593038 on Build GRCh38
rs781647326 Chr.3: 113593081-113593081 on Build GRCh38
rs757725741 Chr.3: 113593059-113593059 on Build GRCh38
rs769366233 Chr.3: 113592989-113592989 on Build GRCh38
rs142833583 Chr.3: 113593026-113593026 on Build GRCh38
rs371038826 Chr.3: 113593085-113593085 on Build GRCh38
rs377625271 Chr.3: 113593021-113593021 on Build GRCh38

View Full Product Details