Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82043852-82044125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEX3B
Entrez Gene ID
84206
Gene Name
mex-3 RNA binding family member B
Gene Aliases
MEX-3B, RKHD3, RNF195
UniGene
Hs.104744
Gene Chromosome Location
Chr.15: 82041778-82046143 on Build GRCh38
Transcripts
NM_032246.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5705966 c.970G>A p.A324T Chr.15: 82043900-82043900 on Build GRCh38
965659 c.873G>A p.S291S Chr.15: 82043997-82043997 on Build GRCh38
4621420 c.994A>G p.N332D Chr.15: 82043876-82043876 on Build GRCh38
346437 c.853C>T p.R285C Chr.15: 82044017-82044017 on Build GRCh38
1257645 c.906C>T p.G302G Chr.15: 82043964-82043964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930554536 Chr.15: 82043893-82043893 on Build GRCh38
rs768756427 Chr.15: 82043898-82043898 on Build GRCh38
rs867327884 Chr.15: 82044068-82044068 on Build GRCh38
rs771636015 Chr.15: 82044085-82044085 on Build GRCh38
rs748332370 Chr.15: 82043907-82043907 on Build GRCh38
rs1022515704 Chr.15: 82044024-82044024 on Build GRCh38
rs745465397 Chr.15: 82043907-82043908 on Build GRCh38
rs551927180 Chr.15: 82043940-82043940 on Build GRCh38
rs747243722 Chr.15: 82044061-82044061 on Build GRCh38
rs754249266 Chr.15: 82044098-82044098 on Build GRCh38
rs760524100 Chr.15: 82044049-82044065 on Build GRCh38
rs61752776 Chr.15: 82044049-82044049 on Build GRCh38
rs766715811 Chr.15: 82044096-82044096 on Build GRCh38
rs750805518 Chr.15: 82043950-82043950 on Build GRCh38
rs769641811 Chr.15: 82044062-82044062 on Build GRCh38
rs758534563 Chr.15: 82044059-82044059 on Build GRCh38
rs773883388 Chr.15: 82044078-82044078 on Build GRCh38
rs745615618 Chr.15: 82044003-82044003 on Build GRCh38
rs200150320 Chr.15: 82044094-82044094 on Build GRCh38
rs112928798 Chr.15: 82044066-82044066 on Build GRCh38
rs775597666 Chr.15: 82043935-82043935 on Build GRCh38
rs756584954 Chr.15: 82043958-82043958 on Build GRCh38
rs770755069 Chr.15: 82043911-82043911 on Build GRCh38
rs781007730 Chr.15: 82044000-82044000 on Build GRCh38
rs774508431 Chr.15: 82043901-82043901 on Build GRCh38
rs145957755 Chr.15: 82043948-82043948 on Build GRCh38
rs762984712 Chr.15: 82044015-82044015 on Build GRCh38
rs1045920819 Chr.15: 82043937-82043937 on Build GRCh38
rs966110459 Chr.15: 82044093-82044093 on Build GRCh38
rs1040715806 Chr.15: 82044047-82044047 on Build GRCh38
rs761190343 Chr.15: 82044080-82044080 on Build GRCh38
rs138885391 Chr.15: 82044006-82044006 on Build GRCh38
rs780499648 Chr.15: 82043970-82043970 on Build GRCh38
rs756929750 Chr.15: 82043942-82043942 on Build GRCh38
rs1030307093 Chr.15: 82043914-82043914 on Build GRCh38
rs768280563 Chr.15: 82044016-82044016 on Build GRCh38
rs775385249 Chr.15: 82044047-82044047 on Build GRCh38
rs754993067 Chr.15: 82043973-82043973 on Build GRCh38
rs776346883 Chr.15: 82043912-82043912 on Build GRCh38
rs754277699 Chr.15: 82044040-82044040 on Build GRCh38
rs772230468 Chr.15: 82043996-82043996 on Build GRCh38
rs973037088 Chr.15: 82043913-82043913 on Build GRCh38
rs748388548 Chr.15: 82043986-82043986 on Build GRCh38
rs1048931820 Chr.15: 82043906-82043906 on Build GRCh38
rs750649615 Chr.15: 82043987-82043987 on Build GRCh38
rs776706874 Chr.15: 82044089-82044090 on Build GRCh38
rs775506159 Chr.15: 82044012-82044012 on Build GRCh38
rs763707689 Chr.15: 82043941-82043941 on Build GRCh38
rs749228578 Chr.15: 82044072-82044072 on Build GRCh38
rs943717081 Chr.15: 82044041-82044041 on Build GRCh38
rs548210663 Chr.15: 82044027-82044027 on Build GRCh38
rs759493708 Chr.15: 82043922-82043922 on Build GRCh38
rs113103544 Chr.15: 82044063-82044063 on Build GRCh38
rs534406222 Chr.15: 82043953-82043953 on Build GRCh38
rs772794896 Chr.15: 82044092-82044092 on Build GRCh38
rs567965511 Chr.15: 82044044-82044044 on Build GRCh38
rs765302990 Chr.15: 82043923-82043923 on Build GRCh38
rs754365892 Chr.15: 82043972-82043972 on Build GRCh38
rs374194178 Chr.15: 82043976-82043976 on Build GRCh38
rs764168356 Chr.15: 82044059-82044060 on Build GRCh38
rs144427196 Chr.15: 82044046-82044046 on Build GRCh38
rs760980649 Chr.15: 82044036-82044036 on Build GRCh38
rs892762379 Chr.15: 82043965-82043965 on Build GRCh38
rs921930748 Chr.15: 82044102-82044102 on Build GRCh38
rs771768286 Chr.15: 82044041-82044042 on Build GRCh38
rs967309500 Chr.15: 82044031-82044031 on Build GRCh38
rs149327902 Chr.15: 82044025-82044025 on Build GRCh38
rs752757395 Chr.15: 82044055-82044055 on Build GRCh38
rs573922527 Chr.15: 82044039-82044039 on Build GRCh38
rs372605609 Chr.15: 82044060-82044060 on Build GRCh38
rs780065951 Chr.15: 82044065-82044065 on Build GRCh38
rs768528475 Chr.15: 82044075-82044075 on Build GRCh38
rs750123863 Chr.15: 82044028-82044028 on Build GRCh38

View Full Product Details