Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95302152-95302397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PON1
Entrez Gene ID
5444
Gene Name
paraoxonase 1
Gene Aliases
ESA, MVCD5, PON
UniGene
Hs.370995
Gene Chromosome Location
Chr.7: 95298357-95324572 on Build GRCh38
Transcripts
NM_000446.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093331 c.801C>T p.L267L Chr.7: 95302313-95302313 on Build GRCh38
5817115 c.797C>A p.T266N Chr.7: 95302317-95302317 on Build GRCh38
1093329 c.871A>C p.I291L Chr.7: 95302243-95302243 on Build GRCh38
3768578 c.793A>G p.N265D Chr.7: 95302321-95302321 on Build GRCh38
4770780 c.781-10_781-2delATCTTTTAA p.? Chr.7: 95302335-95302343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773059400 Chr.7: 95302308-95302308 on Build GRCh38
rs753949449 Chr.7: 95302340-95302340 on Build GRCh38
rs920725709 Chr.7: 95302300-95302300 on Build GRCh38
rs755094281 Chr.7: 95302182-95302182 on Build GRCh38
rs528330068 Chr.7: 95302295-95302295 on Build GRCh38
rs764433951 Chr.7: 95302331-95302331 on Build GRCh38
rs768335187 Chr.7: 95302288-95302288 on Build GRCh38
rs990091064 Chr.7: 95302229-95302229 on Build GRCh38
rs766399556 Chr.7: 95302313-95302313 on Build GRCh38
rs776506565 Chr.7: 95302234-95302236 on Build GRCh38
rs185623242 Chr.7: 95302209-95302209 on Build GRCh38
rs756619244 Chr.7: 95302367-95302368 on Build GRCh38
rs779211745 Chr.7: 95302350-95302350 on Build GRCh38
rs775037874 Chr.7: 95302262-95302262 on Build GRCh38
rs746228258 Chr.7: 95302361-95302361 on Build GRCh38
rs767794219 Chr.7: 95302343-95302344 on Build GRCh38
rs772626843 Chr.7: 95302254-95302254 on Build GRCh38
rs369422555 Chr.7: 95302271-95302271 on Build GRCh38
rs374692491 Chr.7: 95302347-95302347 on Build GRCh38
rs756223967 Chr.7: 95302329-95302329 on Build GRCh38
rs932138233 Chr.7: 95302311-95302311 on Build GRCh38
rs764878087 Chr.7: 95302305-95302305 on Build GRCh38
rs371803280 Chr.7: 95302312-95302312 on Build GRCh38
rs746496670 Chr.7: 95302273-95302273 on Build GRCh38
rs774147610 Chr.7: 95302191-95302191 on Build GRCh38
rs1006910609 Chr.7: 95302194-95302194 on Build GRCh38
rs199693212 Chr.7: 95302239-95302239 on Build GRCh38
rs767776928 Chr.7: 95302184-95302184 on Build GRCh38
rs757515307 Chr.7: 95302245-95302245 on Build GRCh38
rs746294408 Chr.7: 95302252-95302252 on Build GRCh38
rs752865207 Chr.7: 95302317-95302317 on Build GRCh38
rs368248410 Chr.7: 95302303-95302303 on Build GRCh38
rs756454941 Chr.7: 95302200-95302200 on Build GRCh38
rs148911901 Chr.7: 95302248-95302248 on Build GRCh38
rs758951170 Chr.7: 95302369-95302369 on Build GRCh38
rs750758115 Chr.7: 95302354-95302354 on Build GRCh38
rs928440349 Chr.7: 95302291-95302291 on Build GRCh38
rs374147875 Chr.7: 95302195-95302195 on Build GRCh38
rs766896819 Chr.7: 95302368-95302372 on Build GRCh38

View Full Product Details