Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41835714-41835985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL10
Entrez Gene ID
317719
Gene Name
kelch like family member 10
Gene Aliases
SPGF11
UniGene
Hs.127510
Gene Chromosome Location
Chr.17: 41835685-41848347 on Build GRCh38
Transcripts
XM_011524706.2 NM_052935.4 NR_033464.1 NR_033465.1 XM_006721669.3 XM_006721670.3 XM_011524276.2 XM_011524277.2 XM_017024127.1

Gene Symbol
NT5C3B
Entrez Gene ID
115024
Gene Name
5'-nucleotidase, cytosolic IIIB
Gene Aliases
NT5C3L, cN-IIIB
UniGene
Hs.237536
Gene Chromosome Location
Chr.17: 41825082-41836276 on Build GRCh38
Transcripts
XM_011524706.2 NM_052935.4 NR_033464.1 NR_033465.1 XM_006721669.3 XM_006721670.3 XM_011524276.2 XM_011524277.2 XM_017024127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3517572 c.57C>T p.A19A Chr.17: 41835889-41835889 on Build GRCh38
5342936 c.3G>T p.M1I Chr.17: 41835943-41835943 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148308911 Chr.17: 41835931-41835931 on Build GRCh38
rs781864659 Chr.17: 41835849-41835849 on Build GRCh38
rs781980369 Chr.17: 41835911-41835911 on Build GRCh38
rs782291798 Chr.17: 41835890-41835890 on Build GRCh38
rs138262483 Chr.17: 41835947-41835947 on Build GRCh38
rs782195081 Chr.17: 41835963-41835963 on Build GRCh38
rs910464047 Chr.17: 41835957-41835957 on Build GRCh38
rs537090104 Chr.17: 41835965-41835965 on Build GRCh38
rs782120234 Chr.17: 41835821-41835821 on Build GRCh38
rs368420971 Chr.17: 41835892-41835892 on Build GRCh38
rs781997906 Chr.17: 41835814-41835814 on Build GRCh38
rs782400445 Chr.17: 41835888-41835888 on Build GRCh38
rs782522071 Chr.17: 41835861-41835861 on Build GRCh38
rs782685099 Chr.17: 41835868-41835868 on Build GRCh38
rs782510651 Chr.17: 41835939-41835939 on Build GRCh38
rs943405896 Chr.17: 41835794-41835794 on Build GRCh38
rs782172679 Chr.17: 41835870-41835870 on Build GRCh38
rs782007436 Chr.17: 41835814-41835814 on Build GRCh38
rs910463651 Chr.17: 41835786-41835786 on Build GRCh38
rs782133156 Chr.17: 41835912-41835912 on Build GRCh38
rs141466769 Chr.17: 41835933-41835933 on Build GRCh38
rs896001581 Chr.17: 41835840-41835840 on Build GRCh38
rs782077622 Chr.17: 41835894-41835894 on Build GRCh38
rs782053152 Chr.17: 41835829-41835829 on Build GRCh38
rs1865841 Chr.17: 41835736-41835736 on Build GRCh38
rs373979536 Chr.17: 41835837-41835837 on Build GRCh38
rs782309035 Chr.17: 41835891-41835891 on Build GRCh38
rs782582188 Chr.17: 41835961-41835961 on Build GRCh38
rs782808913 Chr.17: 41835822-41835822 on Build GRCh38
rs150436691 Chr.17: 41835934-41835934 on Build GRCh38
rs782128086 Chr.17: 41835821-41835821 on Build GRCh38
rs782555627 Chr.17: 41835950-41835950 on Build GRCh38
rs782713988 Chr.17: 41835831-41835831 on Build GRCh38
rs782020992 Chr.17: 41835889-41835889 on Build GRCh38
rs782788254 Chr.17: 41835914-41835914 on Build GRCh38
rs1009343998 Chr.17: 41835938-41835938 on Build GRCh38
rs782655954 Chr.17: 41835946-41835946 on Build GRCh38
rs782745126 Chr.17: 41835902-41835902 on Build GRCh38
rs547578525 Chr.17: 41835745-41835745 on Build GRCh38
rs782623006 Chr.17: 41835874-41835874 on Build GRCh38
rs782255988 Chr.17: 41835885-41835885 on Build GRCh38
rs899267120 Chr.17: 41835815-41835815 on Build GRCh38
rs565704497 Chr.17: 41835841-41835841 on Build GRCh38
rs781904299 Chr.17: 41835825-41835825 on Build GRCh38
rs929367121 Chr.17: 41835832-41835832 on Build GRCh38
rs782634744 Chr.17: 41835839-41835839 on Build GRCh38
rs781844116 Chr.17: 41835935-41835935 on Build GRCh38

View Full Product Details