Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63546022-63546291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARRES3
Entrez Gene ID
5920
Gene Name
retinoic acid receptor responder 3
Gene Aliases
HRASLS4, HRSL4, PLA1/2-3, RIG1, TIG3
UniGene
Hs.17466
Gene Chromosome Location
Chr.11: 63536801-63546458 on Build GRCh38
Transcripts
NM_004585.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4034983 c.422C>T p.T141M Chr.11: 63546183-63546183 on Build GRCh38
7210349 c.468G>T p.R156S Chr.11: 63546229-63546229 on Build GRCh38
4034982 c.396G>A p.K132K Chr.11: 63546157-63546157 on Build GRCh38
3451295 c.431G>A p.G144E Chr.11: 63546192-63546192 on Build GRCh38
5951367 c.455C>G p.S152C Chr.11: 63546216-63546216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368573715 Chr.11: 63546223-63546223 on Build GRCh38
rs781071378 Chr.11: 63546216-63546216 on Build GRCh38
rs971402720 Chr.11: 63546099-63546099 on Build GRCh38
rs757972293 Chr.11: 63546187-63546187 on Build GRCh38
rs757223365 Chr.11: 63546255-63546255 on Build GRCh38
rs1017280296 Chr.11: 63546068-63546068 on Build GRCh38
rs35845275 Chr.11: 63546246-63546246 on Build GRCh38
rs1024875994 Chr.11: 63546083-63546083 on Build GRCh38
rs746123773 Chr.11: 63546148-63546148 on Build GRCh38
rs764594500 Chr.11: 63546185-63546185 on Build GRCh38
rs764866834 Chr.11: 63546247-63546247 on Build GRCh38
rs754401408 Chr.11: 63546116-63546116 on Build GRCh38
rs761991323 Chr.11: 63546065-63546065 on Build GRCh38
rs752388444 Chr.11: 63546250-63546250 on Build GRCh38
rs75946816 Chr.11: 63546071-63546071 on Build GRCh38
rs1009904448 Chr.11: 63546081-63546081 on Build GRCh38
rs745860467 Chr.11: 63546237-63546237 on Build GRCh38
rs750216038 Chr.11: 63546268-63546268 on Build GRCh38
rs547099153 Chr.11: 63546237-63546237 on Build GRCh38
rs74932327 Chr.11: 63546236-63546236 on Build GRCh38
rs777747960 Chr.11: 63546127-63546127 on Build GRCh38
rs777658252 Chr.11: 63546196-63546196 on Build GRCh38
rs768909251 Chr.11: 63546231-63546231 on Build GRCh38
rs560071292 Chr.11: 63546073-63546073 on Build GRCh38
rs200207046 Chr.11: 63546176-63546176 on Build GRCh38
rs371554112 Chr.11: 63546229-63546229 on Build GRCh38
rs370262815 Chr.11: 63546212-63546212 on Build GRCh38
rs567188777 Chr.11: 63546149-63546149 on Build GRCh38
rs377658071 Chr.11: 63546139-63546139 on Build GRCh38
rs763615313 Chr.11: 63546254-63546254 on Build GRCh38
rs941366824 Chr.11: 63546126-63546126 on Build GRCh38
rs565409700 Chr.11: 63546173-63546173 on Build GRCh38
rs550560829 Chr.11: 63546170-63546170 on Build GRCh38
rs116680849 Chr.11: 63546121-63546121 on Build GRCh38
rs769721435 Chr.11: 63546155-63546155 on Build GRCh38
rs774953457 Chr.11: 63546235-63546235 on Build GRCh38
rs529111789 Chr.11: 63546079-63546079 on Build GRCh38
rs746804956 Chr.11: 63546133-63546133 on Build GRCh38
rs533254357 Chr.11: 63546184-63546184 on Build GRCh38
rs200999428 Chr.11: 63546186-63546186 on Build GRCh38
rs374442728 Chr.11: 63546217-63546217 on Build GRCh38
rs892290057 Chr.11: 63546074-63546074 on Build GRCh38
rs773640180 Chr.11: 63546243-63546243 on Build GRCh38
rs765919879 Chr.11: 63546101-63546101 on Build GRCh38
rs757752183 Chr.11: 63546098-63546098 on Build GRCh38
rs377604818 Chr.11: 63546183-63546183 on Build GRCh38
rs530768706 Chr.11: 63546124-63546124 on Build GRCh38
rs368350490 Chr.11: 63546258-63546258 on Build GRCh38
rs762426234 Chr.11: 63546252-63546252 on Build GRCh38
rs1009375407 Chr.11: 63546267-63546267 on Build GRCh38
rs892313813 Chr.11: 63546264-63546264 on Build GRCh38
rs548784254 Chr.11: 63546119-63546119 on Build GRCh38
rs746006472 Chr.11: 63546222-63546222 on Build GRCh38
rs909380160 Chr.11: 63546169-63546169 on Build GRCh38
rs34628155 Chr.11: 63546115-63546115 on Build GRCh38
rs770798095 Chr.11: 63546136-63546136 on Build GRCh38
rs584905 Chr.11: 63546172-63546172 on Build GRCh38
rs1050146605 Chr.11: 63546047-63546047 on Build GRCh38
rs776552839 Chr.11: 63546236-63546237 on Build GRCh38
rs751545156 Chr.11: 63546199-63546199 on Build GRCh38

View Full Product Details