Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49221024-49221273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLNR
Entrez Gene ID
2862
Gene Name
motilin receptor
Gene Aliases
GPR38, MTLR1
UniGene
Hs.248126
Gene Chromosome Location
Chr.13: 49220338-49222377 on Build GRCh38
Transcripts
NM_001507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047953 c.863G>A p.G288E Chr.13: 49221200-49221200 on Build GRCh38
5512991 c.865C>T p.R289W Chr.13: 49221202-49221202 on Build GRCh38
3957244 c.741C>T p.V247V Chr.13: 49221078-49221078 on Build GRCh38
5754096 c.802G>A p.G268R Chr.13: 49221139-49221139 on Build GRCh38
119706 c.839C>T p.P280L Chr.13: 49221176-49221176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780311565 Chr.13: 49221148-49221149 on Build GRCh38
rs745892813 Chr.13: 49221120-49221121 on Build GRCh38
rs750302335 Chr.13: 49221096-49221096 on Build GRCh38
rs746358130 Chr.13: 49221150-49221150 on Build GRCh38
rs747747480 Chr.13: 49221254-49221254 on Build GRCh38
rs773760214 Chr.13: 49221173-49221173 on Build GRCh38
rs753560677 Chr.13: 49221205-49221205 on Build GRCh38
rs780599909 Chr.13: 49221233-49221233 on Build GRCh38
rs755972660 Chr.13: 49221097-49221097 on Build GRCh38
rs373830822 Chr.13: 49221203-49221203 on Build GRCh38
rs762682548 Chr.13: 49221054-49221054 on Build GRCh38
rs772949328 Chr.13: 49221239-49221240 on Build GRCh38
rs747207076 Chr.13: 49221149-49221169 on Build GRCh38
rs202093803 Chr.13: 49221107-49221107 on Build GRCh38
rs756626390 Chr.13: 49221225-49221225 on Build GRCh38
rs778869981 Chr.13: 49221099-49221101 on Build GRCh38
rs771447627 Chr.13: 49221188-49221188 on Build GRCh38
rs184614513 Chr.13: 49221136-49221136 on Build GRCh38
rs373070788 Chr.13: 49221209-49221209 on Build GRCh38
rs770252835 Chr.13: 49221042-49221042 on Build GRCh38
rs570865470 Chr.13: 49221217-49221217 on Build GRCh38
rs748956186 Chr.13: 49221165-49221165 on Build GRCh38
rs998496207 Chr.13: 49221182-49221182 on Build GRCh38
rs1006681475 Chr.13: 49221065-49221065 on Build GRCh38
rs780511942 Chr.13: 49221164-49221164 on Build GRCh38
rs763192997 Chr.13: 49221211-49221211 on Build GRCh38
rs769124059 Chr.13: 49221244-49221244 on Build GRCh38
rs781743007 Chr.13: 49221148-49221148 on Build GRCh38
rs549854449 Chr.13: 49221210-49221210 on Build GRCh38
rs751701849 Chr.13: 49221213-49221213 on Build GRCh38
rs960336467 Chr.13: 49221047-49221047 on Build GRCh38
rs763714016 Chr.13: 49221062-49221062 on Build GRCh38
rs908238478 Chr.13: 49221076-49221076 on Build GRCh38
rs529713364 Chr.13: 49221239-49221239 on Build GRCh38
rs752971047 Chr.13: 49221100-49221100 on Build GRCh38
rs1050069169 Chr.13: 49221202-49221202 on Build GRCh38
rs749625004 Chr.13: 49221238-49221238 on Build GRCh38
rs757735097 Chr.13: 49221054-49221055 on Build GRCh38
rs750290530 Chr.13: 49221071-49221071 on Build GRCh38
rs777834156 Chr.13: 49221129-49221129 on Build GRCh38
rs781121645 Chr.13: 49221218-49221218 on Build GRCh38
rs200170117 Chr.13: 49221196-49221196 on Build GRCh38
rs772040145 Chr.13: 49221148-49221148 on Build GRCh38
rs939735893 Chr.13: 49221082-49221082 on Build GRCh38
rs760514006 Chr.13: 49221198-49221198 on Build GRCh38
rs778663992 Chr.13: 49221246-49221246 on Build GRCh38
rs146793787 Chr.13: 49221066-49221066 on Build GRCh38
rs868662181 Chr.13: 49221215-49221215 on Build GRCh38
rs761691235 Chr.13: 49221073-49221073 on Build GRCh38
rs538261395 Chr.13: 49221224-49221224 on Build GRCh38
rs766156423 Chr.13: 49221098-49221098 on Build GRCh38
rs1038162876 Chr.13: 49221163-49221163 on Build GRCh38
rs768386901 Chr.13: 49221172-49221172 on Build GRCh38
rs761431105 Chr.13: 49221178-49221178 on Build GRCh38
rs764251471 Chr.13: 49221212-49221212 on Build GRCh38
rs770340952 Chr.13: 49221158-49221158 on Build GRCh38
rs747070715 Chr.13: 49221135-49221135 on Build GRCh38
rs892505258 Chr.13: 49221253-49221253 on Build GRCh38
rs567386610 Chr.13: 49221044-49221044 on Build GRCh38
rs769614097 Chr.13: 49221221-49221221 on Build GRCh38
rs200356464 Chr.13: 49221151-49221151 on Build GRCh38

View Full Product Details