Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68892384-68892657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOX1
Entrez Gene ID
219736
Gene Name
storkhead box 1
Gene Aliases
C10orf24
UniGene
Hs.37636
Gene Chromosome Location
Chr.10: 68827537-68895942 on Build GRCh38
Transcripts
NM_001130159.2 NM_001130160.2 NM_001130161.2 NM_152709.4 XM_011539454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792276 c.2833C>G p.L945V Chr.10: 68892599-68892599 on Build GRCh38
3978738 c.2864G>A p.G955E Chr.10: 68892630-68892630 on Build GRCh38
1561258 c.2836G>A p.G946R Chr.10: 68892602-68892602 on Build GRCh38
919823 c.2884T>G p.F962V Chr.10: 68892650-68892650 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs185349094 Chr.10: 68892589-68892589 on Build GRCh38
rs4472827 Chr.10: 68892516-68892516 on Build GRCh38
rs1014830874 Chr.10: 68892514-68892514 on Build GRCh38
rs765389270 Chr.10: 68892572-68892572 on Build GRCh38
rs758382717 Chr.10: 68892567-68892567 on Build GRCh38
rs757484150 Chr.10: 68892540-68892540 on Build GRCh38
rs758294455 Chr.10: 68892606-68892606 on Build GRCh38
rs751494342 Chr.10: 68892615-68892616 on Build GRCh38
rs763855936 Chr.10: 68892607-68892609 on Build GRCh38
rs142281405 Chr.10: 68892458-68892458 on Build GRCh38
rs386744986 Chr.10: 68892515-68892516 on Build GRCh38
rs76127145 Chr.10: 68892515-68892515 on Build GRCh38
rs752921816 Chr.10: 68892583-68892584 on Build GRCh38
rs545143091 Chr.10: 68892621-68892621 on Build GRCh38
rs769646483 Chr.10: 68892593-68892593 on Build GRCh38
rs758185172 Chr.10: 68892590-68892591 on Build GRCh38
rs151237677 Chr.10: 68892613-68892613 on Build GRCh38
rs757101211 Chr.10: 68892620-68892622 on Build GRCh38
rs993922982 Chr.10: 68892550-68892550 on Build GRCh38
rs745889520 Chr.10: 68892561-68892561 on Build GRCh38
rs781261668 Chr.10: 68892572-68892572 on Build GRCh38
rs189818823 Chr.10: 68892422-68892422 on Build GRCh38
rs777560292 Chr.10: 68892504-68892504 on Build GRCh38
rs772819851 Chr.10: 68892594-68892594 on Build GRCh38
rs10740310 Chr.10: 68892441-68892441 on Build GRCh38
rs748026029 Chr.10: 68892580-68892580 on Build GRCh38
rs750734566 Chr.10: 68892493-68892493 on Build GRCh38
rs1005441834 Chr.10: 68892510-68892510 on Build GRCh38
rs182025494 Chr.10: 68892553-68892553 on Build GRCh38
rs1044279965 Chr.10: 68892494-68892494 on Build GRCh38

View Full Product Details