Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73803010-73803255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDST2
Entrez Gene ID
8509
Gene Name
N-deacetylase and N-sulfotransferase 2
Gene Aliases
HSST2, NST2
UniGene
Hs.654758
Gene Chromosome Location
Chr.10: 73801911-73811831 on Build GRCh38
Transcripts
NM_003635.3 XM_005270255.3 XM_005270256.3 XM_011540310.2 XM_017016857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7107888 c.2331G>T p.G777G Chr.10: 73803064-73803064 on Build GRCh38
5415761 c.2362A>G p.M788V Chr.10: 73803033-73803033 on Build GRCh38
7118763 c.2306C>G p.S769C Chr.10: 73803196-73803196 on Build GRCh38
7107889 c.174G>T p.G58G Chr.10: 73803064-73803064 on Build GRCh38
6723626 c.2353G>A p.A785T Chr.10: 73803042-73803042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051634546 Chr.10: 73803224-73803224 on Build GRCh38
rs762368945 Chr.10: 73803072-73803072 on Build GRCh38
rs755881960 Chr.10: 73803176-73803178 on Build GRCh38
rs754200228 Chr.10: 73803069-73803069 on Build GRCh38
rs527527568 Chr.10: 73803210-73803210 on Build GRCh38
rs757945155 Chr.10: 73803088-73803088 on Build GRCh38
rs759665552 Chr.10: 73803053-73803053 on Build GRCh38
rs769407796 Chr.10: 73803110-73803110 on Build GRCh38
rs371214914 Chr.10: 73803054-73803054 on Build GRCh38
rs777408611 Chr.10: 73803227-73803227 on Build GRCh38
rs772964699 Chr.10: 73803033-73803033 on Build GRCh38
rs764484685 Chr.10: 73803067-73803067 on Build GRCh38
rs765791196 Chr.10: 73803082-73803082 on Build GRCh38
rs763335236 Chr.10: 73803208-73803208 on Build GRCh38
rs938976959 Chr.10: 73803164-73803164 on Build GRCh38
rs928800835 Chr.10: 73803145-73803145 on Build GRCh38
rs373116890 Chr.10: 73803106-73803106 on Build GRCh38
rs750665827 Chr.10: 73803139-73803141 on Build GRCh38
rs760956273 Chr.10: 73803174-73803174 on Build GRCh38
rs376261105 Chr.10: 73803175-73803175 on Build GRCh38
rs776779466 Chr.10: 73803196-73803196 on Build GRCh38
rs866471570 Chr.10: 73803206-73803206 on Build GRCh38
rs775403285 Chr.10: 73803146-73803146 on Build GRCh38
rs767282380 Chr.10: 73803218-73803218 on Build GRCh38
rs779405819 Chr.10: 73803091-73803091 on Build GRCh38
rs140141393 Chr.10: 73803222-73803222 on Build GRCh38
rs370207374 Chr.10: 73803201-73803201 on Build GRCh38
rs199674268 Chr.10: 73803139-73803139 on Build GRCh38
rs374643232 Chr.10: 73803085-73803085 on Build GRCh38
rs754670120 Chr.10: 73803100-73803100 on Build GRCh38
rs373911844 Chr.10: 73803217-73803217 on Build GRCh38
rs749132562 Chr.10: 73803130-73803130 on Build GRCh38
rs760866283 Chr.10: 73803063-73803063 on Build GRCh38
rs373025860 Chr.10: 73803124-73803124 on Build GRCh38
rs762174772 Chr.10: 73803200-73803200 on Build GRCh38
rs767596237 Chr.10: 73803124-73803124 on Build GRCh38
rs781189342 Chr.10: 73803104-73803104 on Build GRCh38
rs752357955 Chr.10: 73803225-73803225 on Build GRCh38
rs751295028 Chr.10: 73803094-73803094 on Build GRCh38
rs777798099 Chr.10: 73803111-73803111 on Build GRCh38
rs143764449 Chr.10: 73803226-73803226 on Build GRCh38

View Full Product Details