Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67661795-67662068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARD6A
Entrez Gene ID
50855
Gene Name
par-6 family cell polarity regulator alpha
Gene Aliases
PAR-6A, PAR6, PAR6C, PAR6alpha, TAX40, TIP-40
UniGene
Hs.112933
Gene Chromosome Location
Chr.16: 67660948-67662778 on Build GRCh38
Transcripts
NM_001037281.1 NM_016948.2 XM_005255977.3 XM_011523095.2 XM_011523096.2 XM_017023261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851248 c.331C>T p.Q111* Chr.16: 67661937-67661937 on Build GRCh38
6737718 c.382C>T p.R128W Chr.16: 67661988-67661988 on Build GRCh38
4642041 c.384G>A p.R128R Chr.16: 67661990-67661990 on Build GRCh38
1168823 c.289_290insGAG p.A96_E97insG Chr.16: 67661895-67661896 on Build GRCh38
3377930 c.358C>T p.R120W Chr.16: 67661964-67661964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746541439 Chr.16: 67661976-67661976 on Build GRCh38
rs763764694 Chr.16: 67662048-67662048 on Build GRCh38
rs766279518 Chr.16: 67662024-67662024 on Build GRCh38
rs774524526 Chr.16: 67661994-67661994 on Build GRCh38
rs746191157 Chr.16: 67661983-67661983 on Build GRCh38
rs781558045 Chr.16: 67661982-67661982 on Build GRCh38
rs144160135 Chr.16: 67661909-67661909 on Build GRCh38
rs1031117174 Chr.16: 67661830-67661830 on Build GRCh38
rs771489203 Chr.16: 67661876-67661877 on Build GRCh38
rs983347310 Chr.16: 67661932-67661932 on Build GRCh38
rs367827389 Chr.16: 67661869-67661869 on Build GRCh38
rs777327701 Chr.16: 67661860-67661860 on Build GRCh38
rs771388310 Chr.16: 67661864-67661864 on Build GRCh38
rs1029329623 Chr.16: 67661990-67661990 on Build GRCh38
rs764264611 Chr.16: 67661900-67661900 on Build GRCh38
rs139574276 Chr.16: 67662029-67662029 on Build GRCh38
rs771555619 Chr.16: 67661850-67661850 on Build GRCh38
rs747479522 Chr.16: 67661862-67661862 on Build GRCh38
rs757994924 Chr.16: 67662047-67662047 on Build GRCh38
rs948813157 Chr.16: 67661841-67661841 on Build GRCh38
rs1024896060 Chr.16: 67661881-67661881 on Build GRCh38
rs775871868 Chr.16: 67661889-67661889 on Build GRCh38
rs761803222 Chr.16: 67662000-67662000 on Build GRCh38
rs1045844851 Chr.16: 67661853-67661853 on Build GRCh38
rs555623024 Chr.16: 67661944-67661944 on Build GRCh38
rs537272237 Chr.16: 67661929-67661929 on Build GRCh38
rs980964562 Chr.16: 67661911-67661911 on Build GRCh38
rs766257662 Chr.16: 67661912-67661912 on Build GRCh38
rs754742811 Chr.16: 67661920-67661920 on Build GRCh38
rs765074184 Chr.16: 67661924-67661924 on Build GRCh38
rs893352969 Chr.16: 67661916-67661916 on Build GRCh38
rs769996618 Chr.16: 67661883-67661883 on Build GRCh38
rs776418775 Chr.16: 67662025-67662025 on Build GRCh38
rs769050563 Chr.16: 67661993-67661993 on Build GRCh38
rs777239811 Chr.16: 67661947-67661947 on Build GRCh38
rs752533462 Chr.16: 67662038-67662038 on Build GRCh38
rs746251847 Chr.16: 67661878-67661878 on Build GRCh38
rs745562541 Chr.16: 67661930-67661931 on Build GRCh38
rs140078781 Chr.16: 67661837-67661837 on Build GRCh38
rs753600700 Chr.16: 67661914-67661914 on Build GRCh38
rs201635871 Chr.16: 67661871-67661871 on Build GRCh38
rs79241223 Chr.16: 67661882-67661882 on Build GRCh38
rs752339028 Chr.16: 67661926-67661926 on Build GRCh38
rs150549185 Chr.16: 67661941-67661941 on Build GRCh38
rs776065821 Chr.16: 67661987-67661987 on Build GRCh38
rs1015529840 Chr.16: 67662036-67662039 on Build GRCh38
rs774843116 Chr.16: 67661890-67661891 on Build GRCh38
rs940383511 Chr.16: 67661972-67661972 on Build GRCh38
rs749490861 Chr.16: 67661989-67661989 on Build GRCh38
rs757745319 Chr.16: 67661977-67661977 on Build GRCh38
rs763325696 Chr.16: 67661892-67661892 on Build GRCh38
rs574032470 Chr.16: 67661984-67661984 on Build GRCh38
rs928661077 Chr.16: 67661877-67661877 on Build GRCh38
rs749692596 Chr.16: 67661875-67661876 on Build GRCh38
rs759583879 Chr.16: 67662026-67662026 on Build GRCh38

View Full Product Details