Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75581236-75581493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLK
Entrez Gene ID
51426
Gene Name
polymerase (DNA) kappa
Gene Aliases
DINB1, DINP, POLQ
UniGene
Hs.135756
Gene Chromosome Location
Chr.5: 75511832-75602576 on Build GRCh38
Transcripts
NM_016218.2 XM_005248534.4 XM_011543463.2 XM_011543464.2 XM_011543467.2 XM_017009558.1 XM_017009559.1 XM_017009560.1 XM_017009561.1 XM_017009562.1 XM_017009563.1 XM_017009564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298676 c.824A>G p.N275S Chr.5: 75581338-75581338 on Build GRCh38
5713502 c.818_821delAACA p.Q274fs*20 Chr.5: 75581332-75581335 on Build GRCh38
5677428 c.919C>G p.L307V Chr.5: 75581433-75581433 on Build GRCh38
4835122 c.727G>T p.E243* Chr.5: 75581241-75581241 on Build GRCh38
25650 c.857T>C p.F286S Chr.5: 75581371-75581371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200548318 Chr.5: 75581406-75581406 on Build GRCh38
rs762382051 Chr.5: 75581323-75581323 on Build GRCh38
rs140423988 Chr.5: 75581417-75581417 on Build GRCh38
rs750829402 Chr.5: 75581383-75581383 on Build GRCh38
rs1044909048 Chr.5: 75581363-75581363 on Build GRCh38
rs35761274 Chr.5: 75581315-75581315 on Build GRCh38
rs368303484 Chr.5: 75581327-75581327 on Build GRCh38
rs145265233 Chr.5: 75581265-75581265 on Build GRCh38
rs148697539 Chr.5: 75581369-75581369 on Build GRCh38
rs759231525 Chr.5: 75581299-75581299 on Build GRCh38
rs934980261 Chr.5: 75581268-75581268 on Build GRCh38
rs5744668 Chr.5: 75581461-75581461 on Build GRCh38
rs771666813 Chr.5: 75581438-75581438 on Build GRCh38
rs142203892 Chr.5: 75581388-75581388 on Build GRCh38
rs779105302 Chr.5: 75581409-75581409 on Build GRCh38
rs372026257 Chr.5: 75581360-75581360 on Build GRCh38
rs1014578762 Chr.5: 75581374-75581374 on Build GRCh38
rs151251843 Chr.5: 75581407-75581407 on Build GRCh38
rs3213801 Chr.5: 75581441-75581441 on Build GRCh38
rs752005211 Chr.5: 75581330-75581330 on Build GRCh38
rs764827018 Chr.5: 75581305-75581305 on Build GRCh38
rs557718700 Chr.5: 75581307-75581307 on Build GRCh38
rs970644341 Chr.5: 75581352-75581352 on Build GRCh38
rs759427560 Chr.5: 75581460-75581460 on Build GRCh38
rs867940075 Chr.5: 75581440-75581440 on Build GRCh38
rs147610161 Chr.5: 75581325-75581325 on Build GRCh38
rs985569860 Chr.5: 75581459-75581459 on Build GRCh38
rs894981954 Chr.5: 75581280-75581280 on Build GRCh38
rs745529078 Chr.5: 75581459-75581460 on Build GRCh38
rs746918425 Chr.5: 75581415-75581415 on Build GRCh38
rs773915529 Chr.5: 75581284-75581286 on Build GRCh38
rs201850011 Chr.5: 75581423-75581423 on Build GRCh38
rs780166089 Chr.5: 75581391-75581391 on Build GRCh38
rs770737000 Chr.5: 75581276-75581277 on Build GRCh38
rs1004979015 Chr.5: 75581358-75581358 on Build GRCh38

View Full Product Details