Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13933065-13933302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
UniGene
Hs.448497
Gene Chromosome Location
Chr.19: 13931187-13953392 on Build GRCh38
Transcripts
NM_001146254.1 NM_001146255.1 NM_024825.3 XM_006722901.1 XM_006722902.1 XM_006722903.1 XM_011528308.1 XM_011528309.2 XM_011528310.2 XM_011528311.1 XM_011528313.1 XM_011528314.1 XM_011528316.1 XM_017027313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611650 c.1072C>T p.R358C Chr.19: 13933172-13933172 on Build GRCh38
4796815 c.995C>T p.A332V Chr.19: 13933249-13933249 on Build GRCh38
6239020 c.1022C>A p.T341K Chr.19: 13933222-13933222 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs10423549 Chr.19: 13933254-13933254 on Build GRCh38
rs749163395 Chr.19: 13933234-13933234 on Build GRCh38
rs375379231 Chr.19: 13933256-13933256 on Build GRCh38
rs1008133197 Chr.19: 13933238-13933238 on Build GRCh38
rs376362504 Chr.19: 13933169-13933169 on Build GRCh38
rs767820910 Chr.19: 13933239-13933239 on Build GRCh38
rs759960334 Chr.19: 13933257-13933257 on Build GRCh38
rs751151283 Chr.19: 13933193-13933193 on Build GRCh38
rs755008910 Chr.19: 13933235-13933235 on Build GRCh38
rs368368090 Chr.19: 13933215-13933215 on Build GRCh38
rs1055371525 Chr.19: 13933137-13933137 on Build GRCh38
rs772160187 Chr.19: 13933243-13933243 on Build GRCh38
rs202013571 Chr.19: 13933168-13933168 on Build GRCh38
rs773385161 Chr.19: 13933249-13933249 on Build GRCh38
rs761936290 Chr.19: 13933160-13933160 on Build GRCh38
rs373561110 Chr.19: 13933230-13933230 on Build GRCh38
rs781131453 Chr.19: 13933117-13933117 on Build GRCh38
rs200487966 Chr.19: 13933136-13933136 on Build GRCh38
rs201236546 Chr.19: 13933129-13933129 on Build GRCh38
rs773715294 Chr.19: 13933280-13933280 on Build GRCh38
rs951089345 Chr.19: 13933162-13933162 on Build GRCh38
rs763503040 Chr.19: 13933192-13933192 on Build GRCh38
rs745701176 Chr.19: 13933118-13933118 on Build GRCh38
rs903294147 Chr.19: 13933221-13933221 on Build GRCh38
rs193206432 Chr.19: 13933228-13933228 on Build GRCh38
rs982627421 Chr.19: 13933163-13933163 on Build GRCh38
rs542625746 Chr.19: 13933104-13933104 on Build GRCh38
rs868695775 Chr.19: 13933262-13933264 on Build GRCh38
rs549358408 Chr.19: 13933269-13933269 on Build GRCh38
rs375402993 Chr.19: 13933099-13933099 on Build GRCh38
rs748307018 Chr.19: 13933240-13933240 on Build GRCh38
rs913595668 Chr.19: 13933112-13933112 on Build GRCh38
rs1055752487 Chr.19: 13933180-13933180 on Build GRCh38
rs999002090 Chr.19: 13933098-13933098 on Build GRCh38
rs915805552 Chr.19: 13933190-13933190 on Build GRCh38
rs756831541 Chr.19: 13933194-13933194 on Build GRCh38
rs903263064 Chr.19: 13933086-13933086 on Build GRCh38
rs79400921 Chr.19: 13933199-13933199 on Build GRCh38
rs963781438 Chr.19: 13933130-13933130 on Build GRCh38
rs781043502 Chr.19: 13933213-13933213 on Build GRCh38
rs768557278 Chr.19: 13933146-13933146 on Build GRCh38
rs373736734 Chr.19: 13933107-13933107 on Build GRCh38
rs981742518 Chr.19: 13933250-13933250 on Build GRCh38
rs755434797 Chr.19: 13933200-13933200 on Build GRCh38
rs997881805 Chr.19: 13933244-13933244 on Build GRCh38
rs760980835 Chr.19: 13933171-13933171 on Build GRCh38
rs936266182 Chr.19: 13933100-13933100 on Build GRCh38
rs750129577 Chr.19: 13933222-13933222 on Build GRCh38
rs371711931 Chr.19: 13933253-13933253 on Build GRCh38
rs922372621 Chr.19: 13933157-13933157 on Build GRCh38
rs749305820 Chr.19: 13933106-13933106 on Build GRCh38
rs766762709 Chr.19: 13933172-13933172 on Build GRCh38
rs888257358 Chr.19: 13933148-13933148 on Build GRCh38
rs369158440 Chr.19: 13933090-13933090 on Build GRCh38
rs774547846 Chr.19: 13933149-13933149 on Build GRCh38
rs562783628 Chr.19: 13933191-13933191 on Build GRCh38
rs754277907 Chr.19: 13933175-13933175 on Build GRCh38
rs371100559 Chr.19: 13933131-13933131 on Build GRCh38
rs754367853 Chr.19: 13933097-13933097 on Build GRCh38

View Full Product Details