Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35746647-35746892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSENEN
Entrez Gene ID
55851
Gene Name
presenilin enhancer gamma-secretase subunit
Gene Aliases
MDS033, MSTP064, PEN-2, PEN2
UniGene
Hs.534465
Gene Chromosome Location
Chr.19: 35745577-35747155 on Build GRCh38
Transcripts
NM_001281532.1 NM_172341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6757477 c.176G>A p.R59H Chr.19: 35746717-35746717 on Build GRCh38
3198673 c.250C>T p.R84C Chr.19: 35746791-35746791 on Build GRCh38
1612097 c.206T>C p.I69T Chr.19: 35746747-35746747 on Build GRCh38
246823 c.167A>C p.Y56S Chr.19: 35746708-35746708 on Build GRCh38
5464498 c.306A>C p.*102C Chr.19: 35746847-35746847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747472843 Chr.19: 35746672-35746672 on Build GRCh38
rs755187533 Chr.19: 35746759-35746759 on Build GRCh38
rs199977740 Chr.19: 35746865-35746865 on Build GRCh38
rs543333097 Chr.19: 35746869-35746869 on Build GRCh38
rs200954908 Chr.19: 35746851-35746851 on Build GRCh38
rs961070064 Chr.19: 35746700-35746700 on Build GRCh38
rs201053747 Chr.19: 35746841-35746841 on Build GRCh38
rs762527582 Chr.19: 35746703-35746703 on Build GRCh38
rs768288547 Chr.19: 35746813-35746813 on Build GRCh38
rs199769543 Chr.19: 35746786-35746786 on Build GRCh38
rs377423067 Chr.19: 35746790-35746790 on Build GRCh38
rs148274462 Chr.19: 35746838-35746838 on Build GRCh38
rs201760819 Chr.19: 35746854-35746854 on Build GRCh38
rs141943005 Chr.19: 35746781-35746781 on Build GRCh38
rs752919606 Chr.19: 35746863-35746863 on Build GRCh38
rs200657188 Chr.19: 35746870-35746870 on Build GRCh38
rs762615142 Chr.19: 35746798-35746798 on Build GRCh38
rs145741019 Chr.19: 35746792-35746792 on Build GRCh38
rs376171447 Chr.19: 35746671-35746671 on Build GRCh38
rs764182589 Chr.19: 35746704-35746704 on Build GRCh38
rs545850323 Chr.19: 35746754-35746754 on Build GRCh38
rs11552441 Chr.19: 35746748-35746748 on Build GRCh38
rs201293694 Chr.19: 35746780-35746780 on Build GRCh38
rs558626989 Chr.19: 35746670-35746670 on Build GRCh38
rs201263466 Chr.19: 35746682-35746682 on Build GRCh38
rs200584017 Chr.19: 35746681-35746681 on Build GRCh38
rs983662386 Chr.19: 35746714-35746714 on Build GRCh38
rs775266605 Chr.19: 35746797-35746797 on Build GRCh38
rs767234224 Chr.19: 35746743-35746743 on Build GRCh38
rs911157410 Chr.19: 35746766-35746766 on Build GRCh38
rs985375925 Chr.19: 35746757-35746757 on Build GRCh38
rs761682700 Chr.19: 35746846-35746846 on Build GRCh38
rs760443048 Chr.19: 35746855-35746855 on Build GRCh38
rs202186580 Chr.19: 35746692-35746692 on Build GRCh38
rs554581670 Chr.19: 35746776-35746776 on Build GRCh38
rs762531363 Chr.19: 35746788-35746788 on Build GRCh38
rs751542345 Chr.19: 35746709-35746709 on Build GRCh38
rs765502200 Chr.19: 35746858-35746858 on Build GRCh38
rs1026598414 Chr.19: 35746736-35746736 on Build GRCh38
rs576399566 Chr.19: 35746674-35746674 on Build GRCh38
rs775176350 Chr.19: 35746702-35746702 on Build GRCh38
rs746209639 Chr.19: 35746791-35746791 on Build GRCh38
rs200199097 Chr.19: 35746697-35746697 on Build GRCh38
rs574419929 Chr.19: 35746787-35746787 on Build GRCh38
rs777455134 Chr.19: 35746869-35746869 on Build GRCh38
rs199535044 Chr.19: 35746848-35746848 on Build GRCh38
rs777852423 Chr.19: 35746867-35746867 on Build GRCh38
rs771521061 Chr.19: 35746865-35746865 on Build GRCh38
rs758461416 Chr.19: 35746782-35746782 on Build GRCh38
rs761745853 Chr.19: 35746725-35746725 on Build GRCh38
rs779075969 Chr.19: 35746772-35746772 on Build GRCh38
rs993882881 Chr.19: 35746716-35746716 on Build GRCh38

View Full Product Details