Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125080619-125080846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A2
Entrez Gene ID
219855
Gene Name
solute carrier family 37 member 2
Gene Aliases
pp11662
UniGene
Hs.352661
Gene Chromosome Location
Chr.11: 125063067-125090516 on Build GRCh38
Transcripts
NM_001145290.1 NM_198277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347523 c.615G>T p.G205G Chr.11: 125080701-125080701 on Build GRCh38
80533 c.597C>A p.A199A Chr.11: 125080683-125080683 on Build GRCh38
924500 c.694C>T p.H232Y Chr.11: 125080780-125080780 on Build GRCh38
7302469 c.657C>T p.A219A Chr.11: 125080743-125080743 on Build GRCh38
1733044 c.613G>A p.G205R Chr.11: 125080699-125080699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371937226 Chr.11: 125080825-125080825 on Build GRCh38
rs34649502 Chr.11: 125080744-125080744 on Build GRCh38
rs991240610 Chr.11: 125080766-125080766 on Build GRCh38
rs754582048 Chr.11: 125080731-125080731 on Build GRCh38
rs200128594 Chr.11: 125080793-125080793 on Build GRCh38
rs377331754 Chr.11: 125080776-125080776 on Build GRCh38
rs954732146 Chr.11: 125080689-125080689 on Build GRCh38
rs368572395 Chr.11: 125080826-125080826 on Build GRCh38
rs780641772 Chr.11: 125080727-125080727 on Build GRCh38
rs370405823 Chr.11: 125080723-125080723 on Build GRCh38
rs749143838 Chr.11: 125080753-125080753 on Build GRCh38
rs201531255 Chr.11: 125080710-125080710 on Build GRCh38
rs372446727 Chr.11: 125080716-125080716 on Build GRCh38
rs769779802 Chr.11: 125080687-125080687 on Build GRCh38
rs758835646 Chr.11: 125080811-125080811 on Build GRCh38
rs531000167 Chr.11: 125080752-125080752 on Build GRCh38
rs371217812 Chr.11: 125080780-125080780 on Build GRCh38
rs943799702 Chr.11: 125080725-125080725 on Build GRCh38
rs553813598 Chr.11: 125080777-125080777 on Build GRCh38
rs761689463 Chr.11: 125080684-125080684 on Build GRCh38
rs144571590 Chr.11: 125080743-125080743 on Build GRCh38
rs368215154 Chr.11: 125080683-125080683 on Build GRCh38
rs140790477 Chr.11: 125080720-125080720 on Build GRCh38
rs765462932 Chr.11: 125080795-125080795 on Build GRCh38
rs773146215 Chr.11: 125080698-125080698 on Build GRCh38
rs779369123 Chr.11: 125080680-125080680 on Build GRCh38
rs780988644 Chr.11: 125080739-125080739 on Build GRCh38
rs760851706 Chr.11: 125080715-125080715 on Build GRCh38
rs770746540 Chr.11: 125080755-125080755 on Build GRCh38
rs374368153 Chr.11: 125080729-125080729 on Build GRCh38
rs376016654 Chr.11: 125080719-125080719 on Build GRCh38
rs755005408 Chr.11: 125080819-125080819 on Build GRCh38
rs750809336 Chr.11: 125080722-125080722 on Build GRCh38
rs918095778 Chr.11: 125080649-125080649 on Build GRCh38
rs752177357 Chr.11: 125080824-125080824 on Build GRCh38
rs772633242 Chr.11: 125080650-125080650 on Build GRCh38
rs536242941 Chr.11: 125080681-125080681 on Build GRCh38
rs750713700 Chr.11: 125080796-125080796 on Build GRCh38
rs200871561 Chr.11: 125080699-125080699 on Build GRCh38
rs1040776101 Chr.11: 125080735-125080735 on Build GRCh38
rs887047183 Chr.11: 125080783-125080783 on Build GRCh38

View Full Product Details