Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40235622-40235881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLF
Entrez Gene ID
6018
Gene Name
rearranged L-myc fusion
Gene Aliases
ZN-15L, ZNF292L
UniGene
Hs.205627
Gene Chromosome Location
Chr.1: 40161369-40240921 on Build GRCh38
Transcripts
NM_012421.3 XM_017002016.1 XM_017002017.1 XM_017002018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063575 c.1173G>T p.K391N Chr.1: 40235875-40235875 on Build GRCh38
681403 c.1178C>G p.S393* Chr.1: 40235880-40235880 on Build GRCh38
4753478 c.1178C>A p.S393* Chr.1: 40235880-40235880 on Build GRCh38
4007899 c.1144C>T p.L382F Chr.1: 40235846-40235846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771314946 Chr.1: 40235672-40235672 on Build GRCh38
rs773689334 Chr.1: 40235767-40235767 on Build GRCh38
rs1016616060 Chr.1: 40235728-40235728 on Build GRCh38
rs184617577 Chr.1: 40235754-40235754 on Build GRCh38
rs544829143 Chr.1: 40235845-40235845 on Build GRCh38
rs982152796 Chr.1: 40235850-40235850 on Build GRCh38
rs763341890 Chr.1: 40235763-40235763 on Build GRCh38
rs759777213 Chr.1: 40235755-40235755 on Build GRCh38
rs111407023 Chr.1: 40235698-40235698 on Build GRCh38
rs774707538 Chr.1: 40235766-40235766 on Build GRCh38
rs778821369 Chr.1: 40235852-40235852 on Build GRCh38
rs1022526290 Chr.1: 40235753-40235753 on Build GRCh38
rs562456117 Chr.1: 40235729-40235729 on Build GRCh38
rs746550604 Chr.1: 40235748-40235748 on Build GRCh38
rs759679286 Chr.1: 40235757-40235758 on Build GRCh38
rs763244361 Chr.1: 40235766-40235767 on Build GRCh38
rs561504935 Chr.1: 40235788-40235788 on Build GRCh38
rs752731441 Chr.1: 40235789-40235789 on Build GRCh38
rs2235985 Chr.1: 40235706-40235706 on Build GRCh38
rs531477120 Chr.1: 40235710-40235714 on Build GRCh38
rs760134779 Chr.1: 40235777-40235777 on Build GRCh38
rs771392869 Chr.1: 40235765-40235765 on Build GRCh38
rs754128889 Chr.1: 40235827-40235827 on Build GRCh38
rs770248608 Chr.1: 40235756-40235757 on Build GRCh38
rs760764033 Chr.1: 40235799-40235799 on Build GRCh38
rs748427076 Chr.1: 40235757-40235757 on Build GRCh38
rs901021711 Chr.1: 40235696-40235697 on Build GRCh38
rs764237211 Chr.1: 40235819-40235819 on Build GRCh38
rs997918944 Chr.1: 40235697-40235698 on Build GRCh38
rs760441744 Chr.1: 40235787-40235787 on Build GRCh38
rs1018079793 Chr.1: 40235669-40235669 on Build GRCh38

View Full Product Details