Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36287102-36287327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THRAP3
Entrez Gene ID
9967
Gene Name
thyroid hormone receptor associated protein 3
Gene Aliases
TRAP150
UniGene
Hs.744057
Gene Chromosome Location
Chr.1: 36224412-36305357 on Build GRCh38
Transcripts
NM_001321471.1 NM_001321473.1 NM_005119.3 XM_005271371.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229819 c.997A>T p.K333* Chr.1: 36287227-36287227 on Build GRCh38
2081218 c.1013C>T p.A338V Chr.1: 36287243-36287243 on Build GRCh38
3419175 c.915T>C p.G305G Chr.1: 36287145-36287145 on Build GRCh38
5629279 c.966_967insT p.S323fs*38 Chr.1: 36287196-36287197 on Build GRCh38
5938785 c.910C>T p.H304Y Chr.1: 36287140-36287140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529988716 Chr.1: 36287294-36287294 on Build GRCh38
rs753730435 Chr.1: 36287214-36287214 on Build GRCh38
rs771220878 Chr.1: 36287266-36287266 on Build GRCh38
rs760698373 Chr.1: 36287193-36287193 on Build GRCh38
rs1034489528 Chr.1: 36287154-36287154 on Build GRCh38
rs138875519 Chr.1: 36287141-36287141 on Build GRCh38
rs780278096 Chr.1: 36287222-36287222 on Build GRCh38
rs751813599 Chr.1: 36287236-36287236 on Build GRCh38
rs1032639069 Chr.1: 36287256-36287256 on Build GRCh38
rs142445892 Chr.1: 36287137-36287137 on Build GRCh38
rs957280921 Chr.1: 36287263-36287263 on Build GRCh38
rs543492113 Chr.1: 36287191-36287191 on Build GRCh38
rs771648723 Chr.1: 36287291-36287291 on Build GRCh38
rs773711351 Chr.1: 36287190-36287190 on Build GRCh38
rs894671087 Chr.1: 36287166-36287166 on Build GRCh38
rs888719019 Chr.1: 36287123-36287123 on Build GRCh38
rs748661109 Chr.1: 36287126-36287126 on Build GRCh38
rs775194797 Chr.1: 36287293-36287293 on Build GRCh38
rs564980038 Chr.1: 36287219-36287219 on Build GRCh38
rs199563213 Chr.1: 36287295-36287295 on Build GRCh38
rs770353571 Chr.1: 36287247-36287247 on Build GRCh38
rs1024499537 Chr.1: 36287217-36287217 on Build GRCh38
rs763780125 Chr.1: 36287207-36287207 on Build GRCh38
rs766096645 Chr.1: 36287179-36287179 on Build GRCh38
rs749711467 Chr.1: 36287261-36287261 on Build GRCh38
rs547439535 Chr.1: 36287274-36287274 on Build GRCh38
rs754986780 Chr.1: 36287238-36287238 on Build GRCh38
rs777969804 Chr.1: 36287254-36287254 on Build GRCh38
rs149836110 Chr.1: 36287287-36287287 on Build GRCh38
rs574148594 Chr.1: 36287208-36287208 on Build GRCh38
rs548216042 Chr.1: 36287296-36287296 on Build GRCh38
rs532099670 Chr.1: 36287243-36287243 on Build GRCh38
rs762508775 Chr.1: 36287165-36287165 on Build GRCh38

View Full Product Details