Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10628526-10628797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH3
Entrez Gene ID
4621
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Gene Aliases
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE
UniGene
Hs.440895
Gene Chromosome Location
Chr.17: 10628526-10679657 on Build GRCh38
Transcripts
NM_002470.3 XM_011523870.2 XM_011523871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7378341 c.5820G>C p.E1940D Chr.17: 10628656-10628656 on Build GRCh38
6892581 c.5818G>A p.E1940K Chr.17: 10628658-10628658 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934575333 Chr.17: 10628712-10628712 on Build GRCh38
rs555639778 Chr.17: 10628661-10628661 on Build GRCh38
rs759613380 Chr.17: 10628699-10628699 on Build GRCh38
rs751631279 Chr.17: 10628696-10628696 on Build GRCh38
rs944325141 Chr.17: 10628741-10628741 on Build GRCh38
rs745429892 Chr.17: 10628656-10628656 on Build GRCh38
rs767666165 Chr.17: 10628707-10628707 on Build GRCh38
rs763494266 Chr.17: 10628692-10628692 on Build GRCh38
rs1054904458 Chr.17: 10628722-10628722 on Build GRCh38
rs529518614 Chr.17: 10628695-10628695 on Build GRCh38
rs534042930 Chr.17: 10628598-10628598 on Build GRCh38
rs955713302 Chr.17: 10628679-10628679 on Build GRCh38
rs757205199 Chr.17: 10628726-10628726 on Build GRCh38
rs755174982 Chr.17: 10628602-10628602 on Build GRCh38
rs758967389 Chr.17: 10628606-10628606 on Build GRCh38
rs780540108 Chr.17: 10628663-10628664 on Build GRCh38
rs371582717 Chr.17: 10628648-10628648 on Build GRCh38
rs777525870 Chr.17: 10628627-10628627 on Build GRCh38
rs915624268 Chr.17: 10628630-10628630 on Build GRCh38
rs912810905 Chr.17: 10628734-10628734 on Build GRCh38
rs752942004 Chr.17: 10628709-10628709 on Build GRCh38
rs201532275 Chr.17: 10628622-10628622 on Build GRCh38
rs749282429 Chr.17: 10628641-10628641 on Build GRCh38
rs770223770 Chr.17: 10628676-10628676 on Build GRCh38
rs753628628 Chr.17: 10628720-10628720 on Build GRCh38
rs959986577 Chr.17: 10628591-10628591 on Build GRCh38
rs775287416 Chr.17: 10628670-10628670 on Build GRCh38
rs781457320 Chr.17: 10628612-10628612 on Build GRCh38
rs747033443 Chr.17: 10628672-10628672 on Build GRCh38
rs770923557 Chr.17: 10628760-10628760 on Build GRCh38
rs886052577 Chr.17: 10628672-10628672 on Build GRCh38
rs373533323 Chr.17: 10628711-10628711 on Build GRCh38
rs368486614 Chr.17: 10628609-10628609 on Build GRCh38
rs770817952 Chr.17: 10628644-10628644 on Build GRCh38
rs982885128 Chr.17: 10628577-10628577 on Build GRCh38
rs372305218 Chr.17: 10628614-10628614 on Build GRCh38
rs777596128 Chr.17: 10628714-10628714 on Build GRCh38
rs1040261672 Chr.17: 10628752-10628752 on Build GRCh38
rs375056974 Chr.17: 10628735-10628735 on Build GRCh38
rs374799651 Chr.17: 10628691-10628691 on Build GRCh38

View Full Product Details